Thermo Scientific™

Owl™ Dual-Gel Vertical Electrophoresis Systems

Catalog number: P9DS-3
Thermo Scientific™

Owl™ Dual-Gel Vertical Electrophoresis Systems

Catalog number: P9DS-3

Quickly and easily cast and run one or two gels in the same device using Thermo Scientific™ Owl™ Dual-Gel Vertical Electrophoresis Systems—comprised of the P8DS, P9DS and P10DS Dual Gel Systems.

 
Catalog Number
P9DS-3
Unit Size
Each
Description
P9DS Apparatus Only
Volume (Metric) Buffer
550 mL
Price (USD)
Price: 1,446.00
Your Price:
Availability
***
Full specifications
Quantity
DescriptionP9DS Apparatus Only
Product LineOwl™
TypeDual-Gel Vertical Electrophoresis System Apparatus
Volume (Metric) Buffer550 mL
Length (English)9.8 in.
Length (Metric) Gel14 to 16 cm
Width (English)5.1 in.
Width (Metric) Gel16 cm
Height (English)7.7 in.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
P10DS Complete System plus Caster
Volume (Metric) Buffer
1250 mL
Price (USD)
Price: 1,981.00
Your Price:
Availability
***
Full specifications
Quantity
DescriptionP10DS Complete System plus Caster
Product LineOwl™
TypeDual-Gel Vertical Electrophoresis Complete System Plus Caster
Gel Dimensions20 x 20  cm
Volume (Metric) Buffer1250 mL
Length (English)12 in.
Length (Metric) Gel20 cm
Width (English)6.5 in.
Width (Metric) Gel20 cm
Height (English)9 in.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
P10DS Complete System
Volume (Metric) Buffer
1250 mL
Price (USD)
Price: 1,949.00
Your Price:
Availability
***
Full specifications
Quantity
DescriptionP10DS Complete System
Product LineOwl™
TypeDual-Gel Vertical Electrophoresis System
Gel Dimensions20 x 20  cm
Volume (Metric) Buffer1250 mL
Length (English)12 in.
Length (Metric) Gel20 cm
Width (English)6.5 in.
Width (Metric) Gel20 cm
Height (English)9 in.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
P10DS Apparatus Only
Volume (Metric) Buffer
1250 mL
Price (USD)
Price: 1,800.00
Your Price:
Availability
***
Full specifications
Quantity
DescriptionP10DS Apparatus Only
Product LineOwl™
TypeDual-Gel Vertical Electrophoresis System Apparatus
Gel Dimensions20 x 20 cm
Volume (Metric) Buffer1250 mL
Length (English)12 in.
Length (Metric) Gel20 cm
Width (English)6.5 in.
Width (Metric) Gel20 cm
Height (English)9 in.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
P8DS Complete System plus Caster
Volume (Metric) Buffer
300 mL
Price (USD)
Price: 1,390.00
Your Price:
Availability
***
Full specifications
Quantity
DescriptionP8DS Complete System plus Caster
Product LineOwl™
TypeDual-Gel Vertical Electrophoresis Complete System with Caster
Gel Dimensions10 x 8-10 cm
Volume (Metric) Buffer300 mL
Length (English)6.7 in.
Length (Metric) Gel8 to 10 cm
Width (English)5.1 in.
Width (Metric) Gel10 cm
Height (English)5.5 in.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
P8DS Complete System
Volume (Metric) Buffer
300 mL
Price (USD)
Price: 1,307.00
Your Price:
Availability
***
Full specifications
Quantity
DescriptionP8DS Complete System
Product LineOwl™
TypeDual-Gel Vertical Electrophoresis System
Gel Dimensions10 x 8-10 cm
Volume (Metric) Buffer300 mL
Length (English)6.7 in.
Length (Metric) Gel8 to 10 cm
Width (English)5.1 in.
Width (Metric) Gel10 cm
Height (English)5.5 in.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
P8DS Apparatus Only
Volume (Metric) Buffer
300 mL
Price (USD)
Price: 1,105.00
Your Price:
Availability
***
Full specifications
Quantity
DescriptionP8DS Apparatus Only
Product LineOwl™
TypeDual-Gel Vertical Electrophoresis System Apparatus
Gel Dimensions10 x 8-10 cm
Volume (Metric) Buffer300 mL
Length (English)6.7 in.
Length (Metric) Gel8 to 10 cm
Width (English)5.1 in.
Width (Metric) Gel10 cm
Height (English)5.5 in.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
P9DS Complete System plus Caster
Volume (Metric) Buffer
550 mL
Price (USD)
Price: 1,655.00
Your Price:
Availability
***
Full specifications
Quantity
DescriptionP9DS Complete System plus Caster
Product LineOwl™
TypeDual-Gel Vertical Electrophoresis Complete System
Volume (Metric) Buffer550 mL
Length (English)9.8 in.
Length (Metric) Gel14 to 16 cm
Width (English)5.1 in.
Width (Metric) Gel16 cm
Height (English)7.7 in.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
P9DS Complete System
Volume (Metric) Buffer
550 mL
Price (USD)
Price: 1,640.00
Your Price:
Availability
***
Full specifications
Quantity
DescriptionP9DS Complete System
Product LineOwl™
TypeDual-Gel Vertical Electrophoresis System
Volume (Metric) Buffer550 mL
Length (English)9.8 in.
Length (Metric) Gel14 to 16 cm
Width (English)5.1 in.
Width (Metric) Gel16 cm
Height (English)7.7 in.
Unit SizeEach
Showing 9 of 9
Catalog NumberSpecificationsUnit SizeDescriptionVolume (Metric) BufferPrice (USD)AvailabilityQuantity
P9DS-3Full specifications
EachP9DS Apparatus Only550 mL
Price: 1,446.00
Your Price:
***
DescriptionP9DS Apparatus Only
Product LineOwl™
TypeDual-Gel Vertical Electrophoresis System Apparatus
Volume (Metric) Buffer550 mL
Length (English)9.8 in.
Length (Metric) Gel14 to 16 cm
Width (English)5.1 in.
Width (Metric) Gel16 cm
Height (English)7.7 in.
Unit SizeEach
P10DS-1Full specifications
EachP10DS Complete System plus Caster1250 mL
Price: 1,981.00
Your Price:
***
DescriptionP10DS Complete System plus Caster
Product LineOwl™
TypeDual-Gel Vertical Electrophoresis Complete System Plus Caster
Gel Dimensions20 x 20  cm
Volume (Metric) Buffer1250 mL
Length (English)12 in.
Length (Metric) Gel20 cm
Width (English)6.5 in.
Width (Metric) Gel20 cm
Height (English)9 in.
Unit SizeEach
P10DS-2Full specifications
EachP10DS Complete System1250 mL
Price: 1,949.00
Your Price:
***
DescriptionP10DS Complete System
Product LineOwl™
TypeDual-Gel Vertical Electrophoresis System
Gel Dimensions20 x 20  cm
Volume (Metric) Buffer1250 mL
Length (English)12 in.
Length (Metric) Gel20 cm
Width (English)6.5 in.
Width (Metric) Gel20 cm
Height (English)9 in.
Unit SizeEach
P10DS-3Full specifications
EachP10DS Apparatus Only1250 mL
Price: 1,800.00
Your Price:
***
DescriptionP10DS Apparatus Only
Product LineOwl™
TypeDual-Gel Vertical Electrophoresis System Apparatus
Gel Dimensions20 x 20 cm
Volume (Metric) Buffer1250 mL
Length (English)12 in.
Length (Metric) Gel20 cm
Width (English)6.5 in.
Width (Metric) Gel20 cm
Height (English)9 in.
Unit SizeEach
P8DS-1Full specifications
EachP8DS Complete System plus Caster300 mL
Price: 1,390.00
Your Price:
***
DescriptionP8DS Complete System plus Caster
Product LineOwl™
TypeDual-Gel Vertical Electrophoresis Complete System with Caster
Gel Dimensions10 x 8-10 cm
Volume (Metric) Buffer300 mL
Length (English)6.7 in.
Length (Metric) Gel8 to 10 cm
Width (English)5.1 in.
Width (Metric) Gel10 cm
Height (English)5.5 in.
Unit SizeEach
P8DS-2Full specifications
EachP8DS Complete System300 mL
Price: 1,307.00
Your Price:
***
DescriptionP8DS Complete System
Product LineOwl™
TypeDual-Gel Vertical Electrophoresis System
Gel Dimensions10 x 8-10 cm
Volume (Metric) Buffer300 mL
Length (English)6.7 in.
Length (Metric) Gel8 to 10 cm
Width (English)5.1 in.
Width (Metric) Gel10 cm
Height (English)5.5 in.
Unit SizeEach
P8DS-3Full specifications
EachP8DS Apparatus Only300 mL
Price: 1,105.00
Your Price:
***
DescriptionP8DS Apparatus Only
Product LineOwl™
TypeDual-Gel Vertical Electrophoresis System Apparatus
Gel Dimensions10 x 8-10 cm
Volume (Metric) Buffer300 mL
Length (English)6.7 in.
Length (Metric) Gel8 to 10 cm
Width (English)5.1 in.
Width (Metric) Gel10 cm
Height (English)5.5 in.
Unit SizeEach
P9DS-1Full specifications
EachP9DS Complete System plus Caster550 mL
Price: 1,655.00
Your Price:
***
DescriptionP9DS Complete System plus Caster
Product LineOwl™
TypeDual-Gel Vertical Electrophoresis Complete System
Volume (Metric) Buffer550 mL
Length (English)9.8 in.
Length (Metric) Gel14 to 16 cm
Width (English)5.1 in.
Width (Metric) Gel16 cm
Height (English)7.7 in.
Unit SizeEach
P9DS-2Full specifications
EachP9DS Complete System550 mL
Price: 1,640.00
Your Price:
***
DescriptionP9DS Complete System
Product LineOwl™
TypeDual-Gel Vertical Electrophoresis System
Volume (Metric) Buffer550 mL
Length (English)9.8 in.
Length (Metric) Gel14 to 16 cm
Width (English)5.1 in.
Width (Metric) Gel16 cm
Height (English)7.7 in.
Unit SizeEach
Showing 9 of 9

Simple, Six-Step Use:

  1. Secure the glass cassettes
  2. Pour the agarose
  3. Plug in the specially designed trough
  4. Cast polyacrylamide solution in the cassettes
  5. Load samples
  6. Run the gel

Features:

  • Manufactured using a time-tested acrylic fabrication technology, providing a durable, long-lasting system
  • Central cooling core provides even cooling across the gel surface for outstanding resolution and clear banding
  • Cooling is achieved by circulating fluid through the core with external circulator or cool tap water
  • Very simple clamping mechanism makes loading glass cassettes and most manufacturers' precast gels trouble free

Recommended for:

  • SDS electrophoresis
  • The second dimension of two-dimensional (2-D) electrophoresis
  • Agarose electrophoresis
  • Native electrophoresis

Figures