Power Blotter Welcome Pack, PVDF
Invitrogen™
Power Blotter Welcome Pack, PVDF
The Invitrogen Power Blotter System (Power Blotter Station + Power Blotter Cassette) is designed specifically for rapid semi-dry transfer ofRead more
Have Questions?
Catalog NumberQuantity
PB01161 Kit
Catalog number PB0116
Price (USD)
4,255.00
Each
Quantity:
1 Kit
Price (USD)
4,255.00
Each
The Invitrogen Power Blotter System (Power Blotter Station + Power Blotter Cassette) is designed specifically for rapid semi-dry transfer of proteins from polyacrylamide gels to nitrocellulose or PVDF membranes in 5 to 12 minutes. The Power Blotter System features an integrated power supply optimized to enable consistent and high-efficiency protein transfer.
Learn more about semi-dry transfer with the Power Blotter System ›
The Power Blotter Welcome Pack, PVDF, includes:
• Power Blotter Station
• Power Blotter Cassette
• Power Blotter Select Transfer Stacks, PVDF, regular size
• Power Blotter Pre-cut Membranes and Filters, PVDF, regular size
• Power Blotter 1-Step Transfer Buffer
Features of the Power Blotter System include:
Efficient—high transfer efficiency with a broad range of protein sizes compared to conventional semi-dry or wet (tank) transfer methods
Fast—5- to 12-minute transfer when used with Power Blotter Select Transfer Stacks or Power Blotter 1-Step Transfer Buffer
Integrated power supply—seamless operation between control unit and cassette; no need for external power supply
Easy-touch programming—convenient pre-programmed transfer methods, or create, save, and run customized transfer methods
Flexible—simultaneous transfer of 1 or 2 mini-size or 1 midi-size gel using the Power Blotter Cassette
Versatile—compatible with Power Blotter Select Transfer Stacks and Pierce 1-Step Transfer Buffer for rapid blotting programs or Towbin transfer buffer for conventional semi-dry transfer methods
The Power Blotter System achieves highly efficient transfer in seven minutes when used with Power Blotter Select Transfer Stacks or ten minutes with Power Blotter 1-Step Transfer Buffer. The instrument is also effective for standard 30- to 60-minute semi-dry transfer protocols based on traditional buffers.
Related products
Power Blotter XL Welcome Pack
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)Acrylamide Gels
Mode of TransferSemi-Dry
Quantity1 Kit
Shipping ConditionRoom Temperature
MaterialPVDF
Product LinePower Blotter
Unit SizeEach
Contents & Storage
• Power Blotter Station
• Power Blotter Cassette
• Power Blotter Select Transfer Stacks, PVDF, regular-size
• Power Blotter Pre-cut Membranes and Filters, PVDF, regular-size
• Power Blotter 1-Step Transfer Buffer