Pesticide Explorer™
Actual product may vary
Thermo Scientific™
Pesticide Explorer™
Start-to-finish LC-MS workflow solutions designed to increase productivity, reduce costs and address the ever-evolving demands for pesticide residue testing.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeMass Spectrometer Interface
PE-QVFB-03HRAM Screening/QuantitationExploris 120 Mass Spectrometer
PE-OTVF-02HRAM QuantitationExploris 120 Mass Spectrometer
PE-OTVF-01Triple Quadrupole QuantitationTriple Quadrupole Mass Spectrometer
Catalog number PE-QVFB-03
Price (USD)
-
Type:
HRAM Screening/Quantitation
Mass Spectrometer Interface:
Exploris 120 Mass Spectrometer
Discover compelling productivity and efficiency enhancements for laboratories performing pesticide residue testing. Thermo Scientific™ Pesticide Explorer™ is a comprehensive liquid chromatography-mass spectrometry (LC-MS) solution designed for laboratories performing routine quantitation and pesticide residue screening in food matrices.
The solution includes all the workflow components needed—consumables, hardware, software and built-in instrument and data processing methods—pre-configured and tested, from your single, trusted supplier, Thermo Fisher Scientific.
Pesticide Explorer provides start-to-finish workflows tailored to help food monitoring and testing laboratories reduce startup time and cost. It provides compelling productivity and efficiency enhancements for both startup laboratories and laboratories adding new analytical capabilities to address evolving customer and industry demands. Regardless of staff or laboratory expertise, Pesticide Explorer is designed to make it easier for both new and experienced users to obtain reliable, unambiguous, high-quality LC-MS/MS results.
Specifications
Certifications/ComplianceCompliant with regulatory methodology (FDA, USDA, SANCO)
Column TypeThermo Scientific™ Accucore™ aQ C18 Polar Endcapped LC column 100 x 2.1 mm
For Use With (Equipment)LC-MS Systems
IncludesUser Guide, Training Materials
Liquid Chromatography SystemThermo Scientific™ Vanquish™ Flex Binary UHPLC
Mass Spectrometer InterfaceExploris 120 Mass Spectrometer
MethodsQuantitation and Screening methods: > 500 pesticides or more (DIA or FSddMS2)
Sample PreparationThermo Scientific™ QuEChERS start-up kit
SoftwareThermo Scientific TraceFinder Software, Thermo Scientific Compound Discoverer Software including mzVault with mzCloud local database
TypeHRAM Screening/Quantitation
Unit SizeEach