PathoProof™ Mastitis Major-3 Kits
PathoProof™ Mastitis Major-3 Kits
Thermo Scientific™

PathoProof™ Mastitis Major-3 Kits

Identify three of the main contagious mastitis-causing microorganisms from bovine milk in just four hours using real-time PCR.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionNo. of Tests
PF0914SPathoProof Mastitis Major - 3 Kit50 Tests
PF0914LPathoProof Mastitis Major - 3 Kit384 Tests
PF0914LBqPCR reagents for use with PF0914L and PF0914LKF384 Tests
PF0914SBqPCR reagents for use with PF0914S and PF0914SKF50 Tests
Catalog number PF0914S
Price (MXN)
-
Description:
PathoProof Mastitis Major - 3 Kit
No. of Tests:
50 Tests
Accurately and rapidly identify three of the main contagious mastitis-causing organisms using Thermo Scientific™ PathoProof™ Mastitis Major-3 Kits. Obtain results within four hours and identify the particular pathogen causing the infection by testing either fresh or preserved milk samples. Make faster decisions on treatment and reduce the time that an infective agent has to spread in a herd.

Identify three of the main contagious mastitis-causing organisms within four hours using PathoProof Major-3 Kits for multiplex PCR. All targeted species are detected simultaneously, overcoming the common problem of mixed growth results in bacterial culturing.

Features include:
• Enables faster decisions on treatment
• Reduces time for infective agents to spread to healthy herd mates
• Reduces broad spectrum antibiotic treatment
• Shortens inflammation time due to rapid treatment
• Enables affected cows to be removed from the herd to ensure optimal milk quality

Assays are based on real-time PCR, an accurate and sensitive technology for identification of the target DNA in the sample. Tests can be used for analyzing fresh and preserved quarter milk samples, milk recording samples and bulk tank milk samples.

Achieve high sensitivity,even for samples that show 'no growth in culture. Accurate detection of viable, dead and growth-inhibited microorganisms reduces “no-growth” results.

PathoProof Major-3 Kit identifies:
1. Mycoplasma bovis
2. Staphylococcus aureus
3. Streptococcus agalactiae

Automated Sample Preparation
Significantly reduce the hands-on time needed when processing larger numbers of samples with the semi-automated Thermo Scientific™ KingFisher™ instrument for magnetic bead-based DNA purification and extraction, along with PathoProof Major-3 KingFisher Kits and qPCR reagents.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Permissible use: This product is intended to be used for the purpose of detection and/or analysis of microorganisms in milk for quality assurance and quality control purposes (Food Testing Applications), as well as for identification, enumeration, or count of microorganisms in raw material sample, process control sample, or finished product sample of an industrial process for the purpose of detecting the presence, absence or amount either of a contaminant or of an intended component (Industrial Microbiology Applications).
Specifications
DescriptionPathoProof Mastitis Major - 3 Kit
For Use With (Equipment)Must be ordered together with PF0914SB qPCR Reagents.
Kit TypeMajor 3
No. of Tests50 Tests
Product TypeReal Time PCR Reagents
For Use With (Application)Food & Beverage
GC-Rich PCR PerformanceMedium
PCR MethodqPCR
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
All necessary reagents (except ethanol) for DNA extraction from milk. Varies by component; qPCR reagents should be stored at –20°C protected from light.