Search
Search

| Catalog Number | Description | For Use With (Equipment) | No. of Tests |
|---|---|---|---|
| PF0942L | PathoProof Mastitis Major – 4.2 Kit | Must be ordered together with PF0942LB qPCR Reagents. | 384 Tests |
| PF0942LB | qPCR reagents for use with PF0942L and PF0942LKF | Must be ordered together with PF0942L or PF0942LKF. | 384 Tests |
| PF0942SB | qPCR reagents for use with PF0942S | Must be ordered together with PF0942S. | 50 Tests |
| PF0942S | PathoProof Mastitis Major – 4.2 Kit | Must be ordered together with PF0942SB qPCR Reagents. | 50 Tests |
PathoProof Mastitis Assays identify the main mastitis-causing microorganisms and the Staphylococcal β-lactamase gene responsible for the penicillin resistance in Staphylococci. These microorganisms are responsible for 95% of all clinical and subclinical mastitis cases worldwide.
• Assays are based on real-time PCR, an accurate and sensitive technology for identification of the target DNA in the sample
• Tests can be used for analyzing fresh and preserved quarter milk samples, milk recording samples and bulk tank milk samples
• All targeted species are detected simultaneously, overcoming the common problem of mixed growth results of bacterial culturing
• Validation studies indicate superior analytical specificity and sensitivity over traditional methods, even for samples that show “no growth” in culture
• Accurate detection of viable, dead and growth-inhibited microorganisms reduces 'no-growth' results
qPCR Reagents for PathoProof Mastitis Complete Assays
• Order item numbers PF1600SB and PF1600LB respectively
• Order item numbers PF0870SB and PF0870LB respectively
qPCR Reagents for PathoProof Mastitis Major Assays
• Order item numbers PF0941SB and PF0941LB respectively
• Order item numbers PF0942SB and PF0942LB respectively
• Order item numbers PF0914SB and PF0914LB respectively
PathoProof qPCR reagents are not sold separately.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.