PathoProof™ Mastitis Complete-16 Kits
Actual product may vary
Thermo Scientific™
PathoProof™ Mastitis Complete-16 Kits
Identify 15 mastitis-causing microorganisms from bovine milk in less than four hours, no culturing required, with Thermo Scientific™ PathoProof™ Mastitis Complete-16 Kit.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionFor Use With (Equipment)No. of Tests
PF1600LBqPCR reagents for use with PF1600L and PF1600LKFMust be ordered together with PF1600L or PF1600LKF.384 Tests
PF1600LKFPathoProof Mastitis Complete-16 KFFor use with the KingFisher Flex system. Must be ordered together with PF1600LB qPCR Reagents.384 Tests
Catalog number PF1600LB
Price (MXN)
-
Description:
qPCR reagents for use with PF1600L and PF1600LKF
For Use With (Equipment):
Must be ordered together with PF1600L or PF1600LKF.
No. of Tests:
384 Tests
Accurately and rapidly identify 15 of the main contagious mastitis-causing organisms and the Staphylococcal β-lactamase gene responsible for penicillin resistance in Staphylococci using Thermo Scientific™ PathoProof™ Mastitis Complete-16 Kits. These microorganisms are responsible for 95% of all clinical and subclinical mastitis cases worldwide. Obtain results within four hours and identify the particular pathogen causing the infection by testing either fresh or preserved milk samples. Make faster decisions on treatment and reduce the time that an infective agent has to spread in a herd.
Identify 15 of the main mastitis-causing microorganisms and the Staphylococcal β-lactamase gene responsible for penicillin resistance in Staphylococci within four hours using PathoProof Complete-16 Kits for multiplex PCR. All targeted species are detected simultaneously, overcoming the common problem of mixed growth results in bacterial culturing.
  • Enables faster decisions on treatment
  • Reduces time for infective agents to spread to healthy herd mates
  • Reduces broad spectrum antibiotic treatment
  • Shortens inflammation time due to rapid treatment
  • Enables affected cows to be removed from the herd to ensure optimal milk quality
Assays are based on real-time PCR, an accurate and sensitive technology for identification of the target DNA in the sample. Tests can be used for analyzing fresh and preserved quarter milk samples, milk recording samples and bulk tank milk samples.
Achieve high sensitivity,even for samples that show no growth in culture. Accurate detection of viable, dead and growth-inhibited microorganisms reduces no-growth results.
PathoProof Mastitis Complete-16 Kit identifies:
  1. Staphylococcus aureus
  2. Staphylococcus spp. (including all major coagulase-negative staphylococci)
  3. Streptococcus agalactiae
  4. Streptococcus dysgalactiae
  5. Streptococcus uberis
  6. Escherichia coli
  7. Enterococcus spp. (including E. faecalis and E. faecium)
  8. Klebsiella oxytoca and K. pneumoniae
  9. Serratia marcescens
  10. Corynebacterium bovis
  11. Arcanobacter pyogenes and Peptoniphilus (Peptostreptococcus) indolicus
  12. Staphylococcal β-lactamase gene (penicillin-resistance gene)
  13. Mycoplasma bovis
  14. Mycoplasma spp.
  15. Yeast
  16. Prototheca spp
Automated sample preparation
Significantly reduce the hands-on time needed when processing larger numbers of samples with the semi-automated Thermo Scientific™ KingFisher™ instrument for magnetic bead-based DNA purification and extraction, along with PathoProof Mastitis Complete-16 KingFisher Kits and qPCR reagents.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Permissible use: This product is intended to be used for the purpose of detection and/or analysis of microorganisms in milk for quality assurance and quality control purposes (Food Testing Applications), as well as for identification, enumeration, or count of microorganisms in raw material sample, process control sample, or finished product sample of an industrial process for the purpose of detecting the presence, absence or amount either of a contaminant or of an intended component (Industrial Microbiology Applications).
Specifications
DescriptionqPCR reagents for use with PF1600L and PF1600LKF
For Use With (Equipment)Must be ordered together with PF1600L or PF1600LKF.
Kit TypeMastitis Kit
No. of Tests384 Tests
Product TypeReal Time PCR Reagents
For Use With (Application)Food & Beverage
PCR MethodqPCR
Unit SizeEach
Contents & Storage
qPCR reagents for use with PF1600L and PF1600LKF