PathoProof™ Mycoplasma-8 Kits for Agilent™ Mx3005P™ qPCR Systems
Actual product may vary
Thermo Scientific™

PathoProof™ Mycoplasma-8 Kits for Agilent™ Mx3005P™ qPCR Systems

Identify mastitis-causing Mycoplasma species from bovine milk in less than four hours, no bacterial culturing required, with Thermo Scientific™ PathoProof™ Mycoplasma - 8 Kits.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionFor Use With (Equipment)No. of Tests
PF8500SPathoProof Mycoplasma – 8 kitFor use with Agilent Mx3005qPCR system. Must be ordered together with PF8500SB qPCR Reagents.50 Tests
PF8500SBPathoProof Mycoplasma – 8 qPCR ReagentsFor use with Agilent Mx3005qPCR system. Must be ordered together with PF8500S.50 Tests
PF8500LPathoProof Mycoplasma – 8 kitFor use with Agilent Mx3005qPCR system. Must be ordered together with PF8500LB qPCR Reagents.384 Tests
PF8500LKFPathoProof Mycoplasma – 8 kitFor use with the KingFisher Flex DNA extraction system and Agilent Mx3005qPCR system. Must be ordered together with PF8500LB qPCR Reagents.384 Tests
PF8500LBPathoProof Mycoplasma – 8 qPCR ReagentsFor use with Agilent Mx3005qPCR system. Must be ordered together with PF8500L or PF8500LKF.384 Tests
Catalog number PF8500S
Price (EUR)
-
Description:
PathoProof Mycoplasma – 8 kit
For Use With (Equipment):
For use with Agilent Mx3005qPCR system. Must be ordered together with PF8500SB qPCR Reagents.
No. of Tests:
50 Tests
Accurately and rapidly identify mastitis-causing Mycoplasma species in bovine milk samples in less than four hours with PathoProof™ Mycoplasma-8 Kit PCR test. The primary benefits of this kit are the time to result (hours compared with 10 days with culture methods), the ability to identify the particular Mycoplasma species causing the infection, and the ability to test both fresh and preserved milk samples. Make faster decisions on treatment and reduce the time that an infective agent has to spread in a herd.

Identify the six main Mycoplasma species together with Staphylococcus aureus and Streptococcus agalactiae in four hours using PathoProof Mycoplasma-8 multiplex PCR. All targeted species are detected simultaneously, overcoming the common problem of mixed growth results in bacterial culturing.

  • Enables faster decisions on treatment
  • Reduces time for infective agents to spread to healthy herd mates
  • Reduces broad spectrum antibiotic treatment
  • Shortens inflammation time
  • Improves milk quality

Assays are based on real-time PCR, an accurate and sensitive technology for identification of the target DNA in the sample. Tests can be used for analyzing fresh and preserved quarter milk samples, milk recording samples and bulk tank milk samples. Achieve high sensitivity, even for samples that show no growth in culture. Accurate detection of viable, dead and growth-inhibited microorganisms reduces no-growth results.

PathoProof Mycoplasma - 8 Kit identifies:

  • Mycoplasma alkalescens
  • Mycoplasma bovis
  • Mycoplasma bovigenitalium
  • Mycoplasma canadense
  • Mycoplasma californicum
  • Mycoplasma spp.
  • Staphylococcus aureus
  • Streptococcus agalactiae

Not all products are available for sale in all territories. Please inquire.

Permissible Use: This product is intended to be used for the purpose of detection and/or analysis of microorganisms in milk for quality assurance and quality control purposes (Food Testing Applications), as well as for identification, enumeration, or count of microorganisms in raw material sample, process control sample, or finished product sample of an industrial process for the purpose of detecting the presence, absence or amount either of a contaminant or of an intended component (Industrial Microbiology Applications).
Specifications
DescriptionPathoProof Mycoplasma – 8 kit
For Use With (Equipment)For use with Agilent Mx3005qPCR system. Must be ordered together with PF8500SB qPCR Reagents.
Kit TypeMycoplasma 8
No. of Tests50 Tests
Product TypeqPCR Reagents for Microbial Detection
Detection MethodqPCR
GC-Rich PCR PerformanceHigh
PCR MethodMultiplex qPCR
Reaction SpeedFast
Unit SizeEach
Contents & Storage
All necessary reagents (except ethanol) for DNA extraction from milk