Thermo Scientific™

Pierce™ Premium Grade Sulfo NHS-SS-Biotin

Catalog number:  PG82078

 Related applications:

Protein Labeling & Crosslinking

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific™ Pierce™ Premium Grade Sulfo-NHS-SS-Biotin is our highest quality formulation of this amine-reactive, cleavable biotinylation reagent, specially characterized for applications where product integrity and risk minimization are paramount. Sulfo-NHS-SS-biotin is used for cleavable attachment of biotin to protein and cell surface amines.

• High quality—identity and purity confirmed by multiple tests, including quantitative NMR
• Product integrity—enhanced level of testing and characterization compared to standard grade
• Lot retention—ample supply of past lots retained to help ensure future process testing
• Change management—Change Control Notification (CCN) service
• Consistent manufacture—batch-specific manufacturing documentation review

Compared to the standard grade product, Premium Grade Sulfo-NHS-SS-Biotin provides more clearly defined quality and product support by including increased analytical testing and product characterization, greater batch-specific information and quality assurance review, extensive lot sample retention, and change control notification.

Sulfo-NHS-SS-biotin is a water-soluble, NHS-ester biotinylation reagent with a spacer arm that includes a cleavable disulfide bond for reversible labeling of proteins and cell surface primary amines. This reagent is water-soluble, enabling biotinylation in the absence of organic solvents such as DMSO or DMF. The compound is particularly useful for labeling and purifying cell surface proteins because its sulfonate group prevents it from permeating cell membranes and its cleavable spacer arm enables initially biotinylated proteins to be released from streptavidin affinity columns.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Permeability: Cell-Impermeant
Spacer: Cleavable, Mid-Length
Label or Dye: Biotin
Chemical Reactivity: Amine
Product Line: Pierce™
Product Size: 1 g
Solubility: DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water

Contents & storage

Premium Grade Sulfo-NHS ester of biotin with an ethyl-1,3-dithioproprionate spacer.
Store with desiccant at -20°C.
Sufficient to label a total of 10 g of IgG using typical conditions.

Documents

Manuals & protocols