Pierce™ Premium Grade DSP (dithiobis(succinimidyl propionate))
Thermo Scientific™

Pierce™ Premium Grade DSP (dithiobis(succinimidyl propionate))

Optimize applications where product integrity and risk minimization are paramount with our highest quality formulation of this popular amine-reactive, cleavable crosslinker.
Have Questions?
產品號碼Quantity
PG820811 g
產品號碼 PG82081
價格 (TWD)
***
Quantity:
1 g
Request bulk or custom format
價格 (TWD)
***

Thermo Scientific Pierce Premium Grade DSP is our highest quality formulation of this popular amine-reactive, cleavable crosslinker, specially characterized for applications where product integrity and risk minimization are paramount.

Compared to the standard grade product, Premium Grade DSP provides more clearly defined quality and product support by including (a) increased analytical testing and product characterization, (b) greater batch-specific information and quality assurance review, (c) extensive lot sample retention, and (d) change control notification. DSP is dithiobis(succinimidyl propionate), an amine-reactive crosslinker having NHS-ester reactive ends and a cleavable disulfide bond in its spacer arm. DSP contains an amine-reactive N-hydroxysuccinimide (NHS) ester at each end of an 8-carbon spacer arm. NHS esters react with primary amines at pH 7-9 to form stable amide bonds, along with release of the N-hydroxy-succinimide leaving group. Proteins, including antibodies, generally have several primary amines in the side chain of lysine (K) residues and the N-terminus of each polypeptide that are available as targets for NHS-ester crosslinking reagents.

Features of Premium Grade DSP:

  • DSP—a thiol-cleavable, homobifunctional and amine-reactive crosslinking agent
  • High quality—identity and purity confirmed by several tests, including quantitative NMR
  • Product integrity—enhanced level of testing and characterization compared to standard grade
  • Lot retention—ample supply of past lots retained to ensure future process testing
  • Change management—Change Control Notification (CCN) service
  • Consistent manufacture—batch-specific manufacturing documentation review

Applications:

  • Chemical crosslinking of intracellular proteins prior to cell lysis and immunoprecipitation
  • 'Fix' protein interactions to allow identification of weak or transient protein interactions
  • Fix tissues for immunostaining without damaging RNA
  • Protein crosslinking to create bioconjugates via single-step reactions
  • Immobilize proteins onto amine-coated surfaces

Product References

Crosslinker Application Guide -- search for recent literature references for this product

Related Products

DSP (dithiobis(succinimidyl propionate)), Lomant's Reagent (Cat. No. 22585)

For Research Use Only. Not for use in diagnostic procedures.
規格
Cell PermeabilityYes
FormPowder
FormulationDithiobis(succinimidylpropionate); powder solid
Labeling MethodChemical Labeling
Molecular Weight (g/mol)404.42
PEGylatedNo
Product LinePierce
Quantity1 g
Reactive MoietyNHS Ester
Shipping ConditionAmbient
SolubilityDMF, DMSO
Spacer Arm Length12.0 Å
Sufficient For2.47L of typical 1X (1mM) reaction solution
Water SolubleNo
Chemical ReactivityAmine-Amine
CleavableBy Thiols
Crosslinker TypeHomobifunctional
FormatStandard, Single-use, Premium-grade
Product TypeCrosslinker
SpacerMedium (10 to 30 Å)
Unit SizeEach
內容物與存放
Upon receipt store desiccated at 4°C.

常見問答集 (常見問題)

Can you provide the shelf-life for Premium Grade DSP (dithiobis(succinimidyl propionate))?

Premium Grade DSP (dithiobis(succinimidyl propionate)) is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.