Human IL-1 alpha Recombinant Protein
Actual product may vary
Gibco™

Human IL-1 alpha Recombinant Protein

Human IL-1 alpha Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FNRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
PHC00142 μg
PHC0011100 μg
PHC00131 mg
PHC00155 μg
PHC001725 μg
Catalog number PHC0014
Price (USD)
-
Quantity:
2 μg
Human IL-1 alpha Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ClassificationCarrier-Free
Endotoxin Level< 0.1 ng/μg
Expression SystemE. coli
Gene AliasIL-1α
Gene ID (Entrez)3552
Mass10 μg
Molecular Weight (g/mol)18.2 kDa
Product LineGibco
Protein FamilyCytokines & Receptors
Protein FormRecombinant, Ligand
Protein SubtypeInterleukins
Purification MethodMembrane-Filtered
Purity or Quality Grade95 %
Quantity2 μg
Research CategoryCell Cycle & Proliferation, Immunology, Angiogenesis
Shipping ConditionApproved for shipment on Wet or Dry Ice
SourceHuman
For Use With (Application)Cell Culture
FormLyophilized
RecombinantRecombinant
SpeciesHuman
Unit Size2 μg
Contents & Storage
Store in refrigerator (2–8°C).

Frequently asked questions (FAQs)

It looks like the recombinant protein vial I received from you is empty. Is this normal?

Gibco recombinant proteins are frequently formulated without carrier proteins or additives (e.g., BSA, HSA, sucrose, etc.) and no Gibco PeproTech recombinant protein contains a carrier protein. As a result, during lyophilization, the protein product may be deposited on the vial as a thin, and sometimes invisible, film instead of a pellet. The size of the pellet, if any, is not directly related to the quantity of the recombinant protein in the vial. Our quality control procedures assure that each vial contains the correct amount of product.

To ensure complete recovery of protein product, before opening a vial of lyophilized recombinant protein, we recommend centrifuging it in a microcentrifuge for 20-30 seconds to drive any protein that may be lodged in the cap or on the side to the bottom of the vial. After reconstitution, you can confirm the presence of product protein by running a small amount on SDS-PAGE. In general, a protein band with expected size should be visible with as little as 10 ng of protein loaded on an acrylamide gel.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Why does my recombinant protein not show activity in my own experiment?

Assay time is critical. Each assay needs to beoptimized and performed at the peak response time. Different cells may respond differently to a growth factor or cytokine. We suggest repeating our QC assay using same indicator cells as suggested in the manual to see if you can obtain a similar response. In addition, serum may be masking the response. Serum starvation might be needed for certain types of assays.

How should I store the reconstituted recombinant proteins?

Protein solutions are generally not very stable when frozen at low concentration. Upon freeze and thaw, some proteins in the solution may stick to the wall of the container, which results in significant reduction of protein concentration if the starting concentration was low. Therefore, carrier proteins are used to reduce such loss. The most commonly used carrier proteins include bovine serum albumin (BSA), human serum albumin (HSA), or fetal bovine serum (FBS). These carrier proteins are generally used at 0.1% concentration. As a rule of thumb, if the concentration of the recombinant protein is less than 0.5 mg/mL, it is a good idea to add some carrier protein

How do I store my lyophilized recombinant protein?

Lyophilized proteins can typically be stored at 2 to 8 degrees C for several weeks, or stored dessicated at -20 degrees C for long-term storage.

Citations & References (1)

Citations & References
Abstract
Up-regulation of Prostaglandin E2 Synthesis by Interleukin-1beta in Human Orbital Fibroblasts Involves Coordinate Induction of Prostaglandin-Endoperoxide H Synthase-2 and Glutathione-dependent Prostaglandin E2 Synthase Expression.
Authors: Han Rui; Tsui Shanli; Smith Terry J;
Journal:J Biol Chem
PubMed ID:11847219
Prostaglandin E(2) (PGE(2)) production involves the activity of a multistep biosynthetic pathway. The terminal components of this cascade, two PGE(2) synthases (PGES), have very recently been identified as glutathione-dependent proteins. cPGES is cytoplasmic, apparently identical to the hsp90 chaperone, p23, and associates functionally with prostaglandin-endoperoxide H synthase-1 (PGHS-1), the constitutive ... More