STAT1 Recombinant Human Protein

Back to top

Description

Recombinant human STAT1 is phosphorylatable in vitro, using either immunoprecipitated JAK1 or JAK2, or whole cell extracts derived from interferon-gamma-stimulated HEK 293 cells.

Format/Formulation: Protein expressed in E. coli, purity (>95%)
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Mass: 100 µg
Protein Form: Recombinant
Purity or Quality Grade: Carrier-Free
Research Category: Signal Transduction, Immunology
Shipping Condition: Approved for shipment on Wet or Dry Ice
Species: Human
Gene ID (Entrez): 6772
Purification Method: Sequential Chromatography
Purity: 95 %
Protein Subtype: STAT Proteins
Validated Application: Kinase Assay
Protein Family: Transcription Factors
Expression System: E. coli

Contents & storage

Store in refrigerator (2–8°C).