Product Image
Thermo Scientific™
Eutech™ pHTestr 10BNC pH Meter
Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications.
Have Questions?
Catalog NumberRange (pH)
PHTEST10BNC-1 to 15
Catalog number PHTEST10BNC
Price (EUR)
175,50
Online exclusive
195,00
Save 19,50 (10%)
Each
Range (pH):
-1 to 15
Price (EUR)
175,50
Online exclusive
195,00
Save 19,50 (10%)
Each
Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications. Eutech™ pH Testr 10 BNC pocket testers are ideal choices for quick and accurate pH checks and routine applications in laboratories, productions plants, and schools.
Features:
  • Large Display Screen - dual display LCD for enhanced readability, provides comprehensive meter information
  • Waterproof, Dustproof Housing - IP67 rated, maintains the integrity of your tester even in harsh conditions. Plus it floats!
  • User-Selectable Electrodes with BNC connectors - for greater flexibility with choice of electrodes for specific applications
  • Push-Button Calibration with Auto-Buffer Recognition - quick, easy calibration with no calibration mistakes
  • Choice of USA or NIST pH buffer standards with up to 3 points
  • Auto-Power Off saves battery power after non-use
  • Hold Function freezes reading momentarily for easy viewing
  • Self-Diagnostic with message codes for easy troubleshooting
Applications
  • General: Ideal for quick and accurate pH checks in pools and spas, aquariums and hydroponics operations, or anywhere frequent pH testing is required.
  • Industrial: Use for cooling towers, food processing water testing and process/wastewater testing in metal finishing, photo development, printing and chemical industries.
  • Educational: Useful in schools, many laboratory applications and ecology studies.
Specifications
Accuracy±0.1 pH
Accuracy (pH)±0.01
Battery Life>500 hr.
Battery Type4 x 1.5V alkaline button cell batteries
DescriptionWaterproof pHTestr 10 BNC with MTC; BNC connector sensor; ± 0.1 pH resolution (meter only – electrode to be ordered separately)
IncludesLarge Dual Custom Display, Self-diagnostic; Hold function; Auto-Power Off after 8.5 minutes
ModelpHTestr 10BNC
Resolution0.1 pH
Temperature CompensationYes (ATC)
CalibrationAutomatic
Calibration Points3
Display TypeLCD, Dual Display with Battery Indicator, Error Messages/Codes
Range (pH)-1 to 15
Resolution (pH)0.01
Unit SizeEach