AG 1478 (Tyrphostin AG1478)

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top


AG1478 is a potent and selective inhibitor of EGFR. A concentration of 0.25 µM is observed to block EGF-stimulated DNA synthesis in Rat-1 fibroblasts by approximately 75%. This compound also blocks EGF-dependent Src-family kinase activation and p21/Cip1/W

Format/Formulation: Lyophilized
For Research Use Only. Not for use in diagnostic procedures.


Form: Solid
Gene ID: 1956
Sterility: Nonsterile
Product Size: 5 mg
Inhibitor Type: Kinase Inhibitor, Kinase Inhibitor (RTK Inhibitor)
Molecular Weight: 315.8 Da
Inhibitor Properties: Chemical
Specific Inhibitor Target: EGFR (Epidermal growth factor receptor)
Shipping Condition: Dry Ice

Contents & storage

Store in freezer (-5 to -30°C).


Manuals & protocols