AG 1478 (Tyrphostin AG1478)

Catalog number: PHZ1194

Invitrogen™  Related applications: Mammalian Cell Culture | Pharma & Biopharma | Protein Assays and Analysis | Target & Lead Identification & Validation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

AG 1478 (also known as Tyrphostin AG1478) is a potent inhibitor of the epidermal growthfactor (EGF) receptor tyrosine kinase. Other tyrosine kinases, including the closely relatedtyrosine kinase HER2, platelet derived growth factor (PDGF) receptor, and p210Bcr-Abl areinhibited only at much higher AG 1478 concentrations.

Format/Formulation: Off-white solid
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Inhibitor Type: Kinase Inhibitor, Kinase Inhibitor (RTK Inhibitor)
Product Size: 5 mg
Specific Inhibitor Target: EGFR (Epidermal growth factor receptor)
Shipping Condition: Dry Ice

Contents & storage

Store in freezer (-5 to -30°C).

Documents

Manuals & protocols