Product Image
Invitrogen™

Rapamycin

Rapamycin
Have Questions?
Catalog NumberQuantity
PHZ12355 mg
Catalog number PHZ1235
Price (USD)
477.00
Each
Quantity:
5 mg
Price (USD)
477.00
Each

Rapamycin is a member of a family of macrolide immunosuppressants that bind to and inhibit the FK506 binding protein (FKBP) proline rotamase.

Format/Formulation: Lyophilized

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Gene AliasFRAP, FRAP1, FRAP2, RAFT1, RAPT1
Gene ID (Entrez)2475
Gene SymbolMTOR
Molecular Weight (g/mol)914.2
Product TypeRapamycin
Proteinmechanistic target of rapamycin (serine/threonine kinase)
Quantity5 mg
Shipping ConditionWet Ice
FormSolid
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30 °C).