RAF1 Kinase Inhibitor I

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Raf1 Kinase Inhibitor I is a potent inhibitor of c-Raf1. This compound has been assessed with a number of other enzymes, including Cdk1, Cdk2, c-Src, ERK2, MEK, p38 MAPK, Tie2,VEGFR2, and c-fms, and has been found to be >=100-fold more selective for Raf1.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form: Solid
Inhibitor Properties: Chemical
Inhibitor Type: Kinase Inhibitor, Kinase Inhibitor (MAP Kinase Inhibitor)
Molecular Weight: 520.9 Da
Product Size: 1 mg
Purification: HPLC
Specific Inhibitor Target: c-RAF 1
Sterility: Nonsterile
Shipping Condition: Dry Ice

Contents & storage

Store in freezer (-5 to -30°C).

Documents

Manuals & protocols