Autoclavable Bottle, 4 L
Invitrogen™

Autoclavable Bottle, 4 L

The Bigfoot Cell Sorter is designed to streamline the cell sorting workflow and enhance the user experience. It has a multitude of cell sorting advancements, accessories, and unique capabilities to enable the most complex multi-parameter sorts, and to meet high-throughput demands for a wide range of applications.
製品番号(カタログ番号)数量
PL003801 x 4 L
製品番号(カタログ番号) PL00380
価格(JPY)
29,000
4 L
数量:
1 x 4 L
The Invitrogen Bigfoot Spectral Cell Sorter provides power, performance, and flexibility to help accelerate cutting edge discoveries. Designed to streamline the cell sorting workflow and enhance the user experience, the Bigfoot sorter has a multitude of cell sorting advancements, accessories, and unique capabilities to enable the most complex multi-parameter sorts, and to meet high-throughput demands for a wide range of applications.
The Invitrogen Bigfoot Spectral Cell Sorter offers a full line of accessory products for continuous expansion of the instrument including interchangeable nozzle tips, temperature controlled sample input and sort output adapters, and bulk fluid bottles and caps.

研究用にのみ使用できます。

仕様
容量(メートル法)4 L
寸法7.5 x 4 x 11 in.
使用対象 (装置)Bigfoot Spectral Cell Sorter
内容Temp Cap
製品ラインBigfoot Spectral Cell Sorter
製品タイプAutoclavable Bottle
数量1 x 4 L
Unit Size4 L
組成および保存条件
Room temperature