Plate Count Agar
Actual product may vary
Plate Count Agar
Plate Count Agar
Thermo Scientific™

Plate Count Agar

Enumerate viable organisms in milk, water, food and dairy products using this non-selective medium.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityFormatMedia TypeManufacturing Origin
PO5013A10/Pk.90mm MonoplatePlate Count AgarGermany
PO0158A10/Pk.90mm MonoplatePlate Count AgarUnited Kingdom
Catalog number PO5013A
Price (KRW)
-
Quantity:
10/Pk.
Format:
90mm Monoplate
Media Type:
Plate Count Agar
Manufacturing Origin:
Germany
Enumerate viable organisms in milk, dairy products, water, food and other specimens of sanitary importance using non-selective Thermo Scientific™ Plate Count Agar.
This standard medium meets the formulation of APHA, AOAC and PHLS.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Certifications/ComplianceTested in accordance with CEN ISO/TS 11133-2.
ColorIvory, transparent
Fill Weight17g ± 5%
Format90mm Monoplate
Media TypePlate Count Agar
Packaging TypeFlow Wrapped - Single Layer
Quantity10/Pk.
Shelf Life14 Weeks
Manufacturing OriginGermany
Product TypeAgar Plate
Unit SizeEach
Contents & Storage
2° to 12°C