Tryptone Soy Agar with Disinhibitor, Contact Plate
Thermo Scientific™

Tryptone Soy Agar with Disinhibitor, Contact Plate

Effortlessly sample surfaces after cleaning and disinfection with a TSA Contact Plate with disinhibitors to neutralize surface disinfectants and aid in microorganism recovery.
Have Questions?
Catalog NumberDescriptionFormatQuantity
PO5024CTryptone Soy Agar with Disinhibitor Contact Plate55mm Contact Plate2 x 10 plates
Catalog number PO5024C
Price (KRW)
-
Description:
Tryptone Soy Agar with Disinhibitor Contact Plate
Format:
55mm Contact Plate
Quantity:
2 x 10 plates
Effortlessly sample surfaces after cleaning and disinfection using Thermo Scientific™ Tryptone Soy Agar with Disinhibitor Contact Plates, containing Lecithin and polysorbate 80, to neutralize surface disinfectants and aid in microorganism recovery.
Prevent agar from falling out of the dish with Thermo Scientific Contact Plates, which feature a dome-shaped bottom. GripFit™ technology makes plates easy-to-handle and ensures the lid remains in place during handling, while being easily removed during environmental monitoring. An inner layer on the base of the plate is sub-divided into grids to allow easy calculation of colony forming units (CFU) from surface tests, enabling labs to report whether contaminant levels fall within pre-defined acceptable limits. Easy stacking provides more stability during handling and transport, and ventilation is reduced to prevent plates from drying out.

Our contact plates are manufactured using a specialized, automated procedure to ensure a smooth, evenly distributed layer of agar on each plate. Surface sampling is achieved by gently rolling the domed surface of the agar onto the area to be tested. If any microorganisms that can cause disease or infection are present, they will grow to form easily visible colonies on the surface of the agar. This offers a reliable and effective way of determining the microbial cleanliness of surface areas and help control invisible dangers associated with microbial contamination. Clearly visible growth is available within three days for bacteria and within five days for fungi.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Certifications/ComplianceTested in accordance with BP/EP/JP/USP.
ColorIvory, transparent
DescriptionTryptone Soy Agar with Disinhibitor Contact Plate
Fill Weight13.5g ± 5%
Format55mm Contact Plate
Packaging TypeFlow Wrapped - Single Layer
Quantity2 x 10 plates
Shelf Life16 Weeks
Manufacturing OriginGermany
Product TypeAgar
Unit SizeEach
Contents & Storage
2 to 25°C