Malt Extract Agar
Thermo Scientific™

Malt Extract Agar

Detect, isolate and enumerate yeasts and molds with this ready-to-use medium.
Have Questions?
Catalog number PO5055A
Price (EUR)
-

Detect, isolate and enumerate yeasts and molds with ready-to-use Thermo Scientific™ Oxoid™ Malt Extract Agar. Mycological peptone in the medium gives a luxuriant growth of yeasts and molds with typical morphology and pigmentation. Due to the acidic nature of the medium, it inhibits the growth of bacteria resulting in improved isolation of yeasts and molds from materials heavily infected with bacteria.

Use Malt Extract Agar for detection, isolation and enumeration of yeasts and molds.

  • Supports luxuriant growth of yeasts and molds while inhibiting the growth of bacteria.

Ready-to-use convenience of prepared media. Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Specifications
ColorOcher yellow, transparent
DescriptionMalt Extract Agar
Fill Weight17g ± 5 %
Format90mm Plate
Packaging TypeFlow Wrapped - Single Layer
Quantity10/Pk.
Shelf Life16 Weeks
Manufacturing OriginGermany
Product TypeAgar Plate
Unit SizeEach
Contents & Storage
2 to 25°C