Brilliance™ ESBL Agar
Thermo Scientific™

Brilliance™ ESBL Agar

Screen clinical samples for the presence of extended-spectrum beta-lactamase (ESBL)-producing bacteria on this novel formulation which uses color to clearly differentiate E.coli.
Have Questions?
Catalog NumberDescriptiondimFormatQuantity
PO5302ABrilliance ESBL Agar10/Pk.
Catalog number PO5302A
Price (EUR)
-
Description:
Brilliance ESBL Agar
Quantity:
10/Pk.
Detect extended-spectrum beta-lactamase-producing Enterobacteriaceae while selectively inhibiting AmpC resistance mechanisms using Thermo Scientific™ Brilliance™ ESBL Agar. Improve processing time through the direct inoculation from screening swab, isolated colony or from a liquid suspension.
ESBL-producing E. coli grow as either blue or pink colonies. ESBL-producing members of the KESC group produce green colonies; Proteus, Morganella and Providencia do not utilize either chromogen but are able to deaminate tryptohan, resulting in tan colonies with a brown halo.

Brilliance ESBL Agar is a selective medium for the screening of clinical samples for the presence of extended-spectrum beta-lactamase (ESBL) producing bacteria. Easily and clearly differentiate E.coli and the KESC group by different colors to help to identify ESBL producing organisms . The inhibition of AmpC producers reduce false positives and the need for confirmatory tests.

This product may not be available in all countries or territories. Please consult your local representative for availability.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Specifications
ColorOyster white, opaque
DescriptionBrilliance ESBL Agar
Fill Weight17g ± 5%
Format90mm Monoplate
Packaging TypeFlow Wrapped - Single Layer
Quantity10/Pk.
Shelf Life8 Weeks
Manufacturing OriginGermany
Product TypeAgar Plate
Unit SizeEach
Contents & Storage
2° to 12°C, in the dark