Product Image
Applied Biosystems™
Pathatrix™ 10 Pooling Kit
Have Questions?
Número de catálogoCantidad
POOL1010MLN1 kit
Número de catálogo POOL1010MLN
Precio (MXN)
-
Cantidad:
1 kit
The Pathatrix® 10 Pooling Kit simplifies the measurement and pooling of samples from 10 individual pre-enriched food and environmental samples. The single-use sterile straws maintain pre-enriched sample integrity for the subsequent analysis of any positive sample. Each kit includes 100 sterile 10 ml syringes and 1000 sterile 254 mm straws combined into convenient packs of 50. Use this kit with the Pathatrix® Foam Filters (available separately) to remove particulate matter from samples prior to pooling.
Especificaciones
Para utilizar con (equipo)Pathatrix™ Auto System
N.º por caja1000 x 250 mm Straws, 100 x 10 mL Syringes
Línea de productosPathatrix
Tipo de productoKit de combinación
Cantidad1 kit
Condiciones de envíoTemperatura ambiente
Unit SizeEach
Contenido y almacenamiento
1000 sterile 254 mm straws in packs of 50.
100 sterile 10 ml syringes in packs of 50.
Store at room temperature.