Thermo Scientific™

Pre-Preserved Environmental Sample Containers

Catalog number: PP140-40CEP.5HA
Thermo Scientific™

Pre-Preserved Environmental Sample Containers

Catalog number: PP140-40CEP.5HA

Simplify field preservation with Thermo Scientific™ Chemically-Preserved Environmental Sample Containers and enjoy peace of mind as well as convenience. Using certified containers and chemicals reduces the risk of container and preservative contamination. Let us save you the time, tedium and chemical contact associated with diluting and aliquoting chemical preservatives to prepare environmental sample container sets. Choose from our standard offering of chemically-preserved containers, or let us custom-create container/preservative combos according to your personal analytical needs.

 
Catalog Number
PP140-40CEP.5HA
Unit Size
Each
Volume (Metric)
40 mL
Color
Clear
Preservative
0.5 mL 1:1 Hydrochloric Acid
Price (USD)
Full specifications
Diameter (English) Outer1.1 in.
ColorClear
Diameter (Metric) Outer28 mm
MaterialGlass (Type 1)
Volume (Metric)40 mL
Height (English)3.75 in.
LevelCertified
No. per Case72
Volume (English)1 oz.
Volume (Metric) Total40 mL
Closure TypeSepta Cap
Height (Metric)95 mm
Thickness (Metric)3.17 mm
Certifications/ComplianceCertified to meet the EPA Performance Based Specifications for volatile organic compounds; packaged
TypeEnvironmental Sample Container
Preservative0.5 mL 1:1 Hydrochloric Acid
Closure Size24-414
Unit SizeEach
Catalog Number
Unit Size
Each
Volume (Metric)
40 mL
Color
Clear
Preservative
0.25 mL 1:1 Hydrochloric Acid
Price (USD)
Full specifications
Diameter (English) Outer1.1 in.
ColorClear
Diameter (Metric) Outer28 mm
MaterialGlass (Type 1)
Volume (Metric)40 mL
Height (English)3.75 in.
LevelCertified
No. per Case72
Volume (English)1 oz.
Volume (Metric) Total40 mL
Closure TypeSepta Cap
Height (Metric)95 mm
Thickness (Metric)3.17 mm
Certifications/ComplianceCertified to meet the EPA Performance Based Specifications for volatile organic compounds; packaged
TypeEnvironmental Sample Container
Preservative0.25 mL 1:1 Hydrochloric Acid
Closure Size24-414
Unit SizeEach
Catalog Number
Unit Size
Each
Volume (Metric)
40 mL
Color
Clear
Preservative
0.2 mL 1:1 Hydrochloric Acid
Price (USD)
Full specifications
Diameter (English) Outer1.1 in.
ColorClear
Diameter (Metric) Outer28 mm
MaterialGlass (Type 1)
Volume (Metric)40 mL
Height (English)3.75 in.
LevelCertified
No. per Case72
Volume (English)1 oz.
Volume (Metric) Total40 mL
Closure TypeSepta Cap
Height (Metric)95 mm
Thickness (Metric)3.17 mm
Certifications/ComplianceCertified to meet the EPA Performance Based Specifications for volatile organic compounds; packaged
TypeEnvironmental Sample Container
Preservative0.2 mL 1:1 Hydrochloric Acid
Closure Size24-414
Unit SizeEach
Catalog Number
Unit Size
Each
Volume (Metric)
40 mL
Color
Amber
Preservative
0.2 mL 1:1 Hydrochloric Acid
Price (USD)
Full specifications
Diameter (English) Outer1.1 in.
ColorAmber
Diameter (Metric) Outer28 mm
MaterialGlass (Type 1)
Volume (Metric)40 mL
Height (English)3.75 in.
LevelCertified
No. per Case72
Volume (English)1 oz.
Volume (Metric) Total40 mL
Closure TypeSepta Cap
Height (Metric)95 mm
Thickness (Metric)3.17 mm
Certifications/ComplianceCertified to meet the EPA Performance Based Specifications for volatile organic compounds; packaged
TypeEnvironmental Sample Container
Preservative0.2 mL 1:1 Hydrochloric Acid
Closure Size24-414
Unit SizeEach
Catalog Number
Unit Size
Each
Volume (Metric)
40 mL
Color
Amber
Preservative
0.5 mL 1:1 Hydrochloric Acid
Price (USD)
Full specifications
Diameter (English) Outer1.1 in.
ColorAmber
Diameter (Metric) Outer28 mm
MaterialGlass (Type 1)
Volume (Metric)40 mL
Height (English)3.75 in.
LevelCertified
No. per Case72
Volume (English)1 oz.
Volume (Metric) Total40 mL
Closure TypeSepta Cap
Height (Metric)95 mm
Thickness (Metric)3.17 mm
Certifications/ComplianceCertified to meet the EPA Performance Based Specifications for volatile organic compounds; packaged
TypeEnvironmental Sample Container
Preservative0.5 mL 1:1 Hydrochloric Acid
Closure Size24-414
Unit SizeEach
Catalog Number
Unit Size
Each
Volume (Metric)
40 mL
Color
Clear
Preservative
5 mL Methanol, tare weighed
Price (USD)
Full specifications
Diameter (English) Outer1.1 in.
ColorClear
Diameter (Metric) Outer28 mm
MaterialGlass (Type 1)
Volume (Metric)40 mL
Height (English)3.75 in.
LevelCertified
No. per Case72
Volume (English)1 oz.
Volume (Metric) Total40 mL
Closure TypeSepta Cap
Height (Metric)95 mm
Thickness (Metric)3.17 mm
Certifications/ComplianceCertified to meet the EPA Performance Based Specifications for volatile organic compounds; packaged
TypeEnvironmental Sample Container
Preservative5 mL Methanol, tare weighed
Closure Size24-414
Unit SizeEach
Catalog Number
Unit Size
Each
Volume (Metric)
40 mL
Color
Clear
Preservative
10 mL Methanol, tare weighed
Price (USD)
Full specifications
Diameter (English) Outer1.1 in.
ColorClear
Diameter (Metric) Outer28 mm
MaterialGlass (Type 1)
Volume (Metric)40 mL
Height (English)3.75 in.
LevelCertified
No. per Case72
Volume (English)1 oz.
Volume (Metric) Total40 mL
Closure TypeSepta Cap
Height (Metric)95 mm
Thickness (Metric)3.17 mm
Certifications/ComplianceCertified to meet the EPA Performance Based Specifications for volatile organic compounds; packaged
TypeEnvironmental Sample Container
Preservative10 mL Methanol, tare weighed
Closure Size24-414
Unit SizeEach
Catalog Number
Unit Size
Each
Volume (Metric)
40 mL
Color
Clear
Preservative
5 mL Sodium Bisulfate, Stir Bar, tare weighed
Price (USD)
Full specifications
Diameter (English) Outer1.1 in.
ColorClear
Diameter (Metric) Outer28 mm
MaterialGlass (Type 1)
Volume (Metric)40 mL
Height (English)3.75 in.
LevelCertified
No. per Case72
Volume (English)1 oz.
Volume (Metric) Total40 mL
Closure TypeSepta Cap
Height (Metric)95 mm
Thickness (Metric)3.17 mm
Certifications/ComplianceCertified to meet the EPA Performance Based Specifications for volatile organic compounds; packaged
TypeEnvironmental Sample Container
Preservative5 mL Sodium Bisulfate, Stir Bar, tare weighed
Closure Size24-414
Unit SizeEach
Showing 8 of 8
Catalog NumberSpecificationsUnit SizeVolume (Metric)ColorPreservativePrice (USD)
PP140-40CEP.5HAFull specifications
Each40 mLClear0.5 mL 1:1 Hydrochloric AcidRequest A Quote
Diameter (English) Outer1.1 in.
ColorClear
Diameter (Metric) Outer28 mm
MaterialGlass (Type 1)
Volume (Metric)40 mL
Height (English)3.75 in.
LevelCertified
No. per Case72
Volume (English)1 oz.
Volume (Metric) Total40 mL
Closure TypeSepta Cap
Height (Metric)95 mm
Thickness (Metric)3.17 mm
Certifications/ComplianceCertified to meet the EPA Performance Based Specifications for volatile organic compounds; packaged
TypeEnvironmental Sample Container
Preservative0.5 mL 1:1 Hydrochloric Acid
Closure Size24-414
Unit SizeEach
PP140-40CEP.25HAFull specifications
Each40 mLClear0.25 mL 1:1 Hydrochloric AcidRequest A Quote
Diameter (English) Outer1.1 in.
ColorClear
Diameter (Metric) Outer28 mm
MaterialGlass (Type 1)
Volume (Metric)40 mL
Height (English)3.75 in.
LevelCertified
No. per Case72
Volume (English)1 oz.
Volume (Metric) Total40 mL
Closure TypeSepta Cap
Height (Metric)95 mm
Thickness (Metric)3.17 mm
Certifications/ComplianceCertified to meet the EPA Performance Based Specifications for volatile organic compounds; packaged
TypeEnvironmental Sample Container
Preservative0.25 mL 1:1 Hydrochloric Acid
Closure Size24-414
Unit SizeEach
PP140-40CEP.2HAFull specifications
Each40 mLClear0.2 mL 1:1 Hydrochloric AcidRequest A Quote
Diameter (English) Outer1.1 in.
ColorClear
Diameter (Metric) Outer28 mm
MaterialGlass (Type 1)
Volume (Metric)40 mL
Height (English)3.75 in.
LevelCertified
No. per Case72
Volume (English)1 oz.
Volume (Metric) Total40 mL
Closure TypeSepta Cap
Height (Metric)95 mm
Thickness (Metric)3.17 mm
Certifications/ComplianceCertified to meet the EPA Performance Based Specifications for volatile organic compounds; packaged
TypeEnvironmental Sample Container
Preservative0.2 mL 1:1 Hydrochloric Acid
Closure Size24-414
Unit SizeEach
PP141-40AEP.2HAFull specifications
Each40 mLAmber0.2 mL 1:1 Hydrochloric AcidRequest A Quote
Diameter (English) Outer1.1 in.
ColorAmber
Diameter (Metric) Outer28 mm
MaterialGlass (Type 1)
Volume (Metric)40 mL
Height (English)3.75 in.
LevelCertified
No. per Case72
Volume (English)1 oz.
Volume (Metric) Total40 mL
Closure TypeSepta Cap
Height (Metric)95 mm
Thickness (Metric)3.17 mm
Certifications/ComplianceCertified to meet the EPA Performance Based Specifications for volatile organic compounds; packaged
TypeEnvironmental Sample Container
Preservative0.2 mL 1:1 Hydrochloric Acid
Closure Size24-414
Unit SizeEach
PP141-40AEP.5HAFull specifications
Each40 mLAmber0.5 mL 1:1 Hydrochloric AcidRequest A Quote
Diameter (English) Outer1.1 in.
ColorAmber
Diameter (Metric) Outer28 mm
MaterialGlass (Type 1)
Volume (Metric)40 mL
Height (English)3.75 in.
LevelCertified
No. per Case72
Volume (English)1 oz.
Volume (Metric) Total40 mL
Closure TypeSepta Cap
Height (Metric)95 mm
Thickness (Metric)3.17 mm
Certifications/ComplianceCertified to meet the EPA Performance Based Specifications for volatile organic compounds; packaged
TypeEnvironmental Sample Container
Preservative0.5 mL 1:1 Hydrochloric Acid
Closure Size24-414
Unit SizeEach
P140-40CEPPTTWFull specifications
Each40 mLClear5 mL Methanol, tare weighedRequest A Quote
Diameter (English) Outer1.1 in.
ColorClear
Diameter (Metric) Outer28 mm
MaterialGlass (Type 1)
Volume (Metric)40 mL
Height (English)3.75 in.
LevelCertified
No. per Case72
Volume (English)1 oz.
Volume (Metric) Total40 mL
Closure TypeSepta Cap
Height (Metric)95 mm
Thickness (Metric)3.17 mm
Certifications/ComplianceCertified to meet the EPA Performance Based Specifications for volatile organic compounds; packaged
TypeEnvironmental Sample Container
Preservative5 mL Methanol, tare weighed
Closure Size24-414
Unit SizeEach
PP140-40CEPPTTWFull specifications
Each40 mLClear10 mL Methanol, tare weighedRequest A Quote
Diameter (English) Outer1.1 in.
ColorClear
Diameter (Metric) Outer28 mm
MaterialGlass (Type 1)
Volume (Metric)40 mL
Height (English)3.75 in.
LevelCertified
No. per Case72
Volume (English)1 oz.
Volume (Metric) Total40 mL
Closure TypeSepta Cap
Height (Metric)95 mm
Thickness (Metric)3.17 mm
Certifications/ComplianceCertified to meet the EPA Performance Based Specifications for volatile organic compounds; packaged
TypeEnvironmental Sample Container
Preservative10 mL Methanol, tare weighed
Closure Size24-414
Unit SizeEach
PP140-40CEPSBTBFull specifications
Each40 mLClear5 mL Sodium Bisulfate, Stir Bar, tare weighedRequest A Quote
Diameter (English) Outer1.1 in.
ColorClear
Diameter (Metric) Outer28 mm
MaterialGlass (Type 1)
Volume (Metric)40 mL
Height (English)3.75 in.
LevelCertified
No. per Case72
Volume (English)1 oz.
Volume (Metric) Total40 mL
Closure TypeSepta Cap
Height (Metric)95 mm
Thickness (Metric)3.17 mm
Certifications/ComplianceCertified to meet the EPA Performance Based Specifications for volatile organic compounds; packaged
TypeEnvironmental Sample Container
Preservative5 mL Sodium Bisulfate, Stir Bar, tare weighed
Closure Size24-414
Unit SizeEach
Showing 8 of 8
  • Standard Thermo Scientific Certified sample containers chemically-preserved with certified chemical preservatives for environmental sample analyses
  • Packaged in compliance with the shipping requirements of 49 CFR Part 173.4 DOT Small Quantities Classification for highway and rail transport

Ordering Information:

Custom preserved containers available by special order:

  • Choose a standard certified Thermo Scientific environmental sample container
  • Choose a chemical from our standard chemical list
  • Specify the volume and concentration of chemical to aliquot into each container

Products available only in the US

Includes:

Certified sample containers dosed with chemical preservative; certificate of analysis for container and preservative, individually barcoded containers traceable to certs, sample analysis labels unattached for optional use, hazardous material compliant packaging. MSDS available upon request.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text