Regulatory requirements vary by country; products may not be available in your geographic area.
PrioCLIP® Container Box (10010)
Catalog number: PR10010
 Related applications: Animal Health
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
The PrioCLIP® Container Box is an extremely robust and economical homogenization container designed exclusively for use with the PrioGENIZER™ Homogenization Device. The PrioCLIP® is user-friendly with an ergonomic shape allowing single-handed manipulation of the lid. The mouth is wide, facilitating the transfer of tissue samples and collection of homogenates.
For Veterinary Use Only.
Specifications
For Use With (Equipment): PR10000 PrioGENIZER®
Product Line: PrioCLIP®, Prionics™
Product Size: 100 pieces
Type: Container Box
Documents
Manuals & protocols