Aluminum Support, tube/plate rack, 384-well
Thermo Scientific™
Aluminum Support, tube/plate rack, 384-well
The Thermo Scientific 384-well Aluminium Support Rack provides optimal support for heat sealing and automated dispensing for all standard 384-wellRead more
Have Questions?
Catalog NumberQuantity
PRK13831 rack
Catalog number PRK1383
Price (USD)
357.78
Special offer
Online exclusive
Ends: 27-Jun-2026
402.00
Save 44.22 (11%)
Each
Quantity:
1 rack
Price (USD)
357.78
Special offer
Online exclusive
Ends: 27-Jun-2026
402.00
Save 44.22 (11%)
Each
The Thermo Scientific 384-well Aluminium Support Rack provides optimal support for heat sealing and automated dispensing for all standard 384-well PCR plates as well as 96-well Piko PCR Plates and Plate/Frame assemblies.
Features
  • V-shaped wells give optimal support for ultra-thin walls for applying and removing sealers
  • Made of blue aluminum
  • Industry standard footprint fits in robotic workstations, enabling automated pipetting with Piko PCR plates
Specifications
For Use With (Equipment)Real-Time PCR Systems or Thermal Cyclers
Quantity1 rack
ColorBlue/Clear, Clear
HoldsMicroplates (384-well)
MaterialAluminum
Product TypeMicroplate Rack
ShapeRectangular
Unit SizeEach