PowerEase™ Touch HV Power Supply Adapters
Invitrogen™
PowerEase™ Touch HV Power Supply Adapters
The Invitrogen PowerEase Touch HV Power Supply Adapters are designed to facilitate the connection of some electrophoresis tanks to theRead more
Have Questions?
Catalog NumberQuantity
PSHV00A1 set
Catalog number PSHV00A
Price (MXN)
-
Quantity:
1 set
The Invitrogen PowerEase Touch HV Power Supply Adapters are designed to facilitate the connection of some electrophoresis tanks to the PowerEase Touch HV Power Supply. Four sets of adapters are included with each HV power supply, but additional sets are offered here for separate purchase.
Supported electrophoresis tanks include:
• ZOOM IPGRunner Mini-cell
• XCell SureLock Mini-Cell
• Mini Gel Tank
• XCell4 SureLock Midi-Cell
The adapters are designed to remain on the electrophoresis tank electrical leads when the lid is removed from the power supply.
The SureLock Tandem Midi Gel Tank does not require these adapters to be compatible with the PowerEase Touch HV power supply.
Learn more about 2D electrophoresis ›
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity1 set
Shipping ConditionRoom Temperature
For Use With (Application)Gel Electrophoresis, Isoelectric Focusing (IEF or IPG)
For Use With (Equipment)XCell SureLock Mini, ZOOM IPGRunner Mini, PowerEase Touch HV Power Supply, XCell4 SureLock Midi, Mini Gel Tank
Product LinePowerEase
Unit SizeEach
Frequently asked questions (FAQs)
Do you offer a power supply that is comparable to the Bio-Rad PowerPac HV Power Supply?
Yes, we recommend using the PowerEase Touch HV Power Supply.
Find additional tips, troubleshooting help, and resources within our Protein Gel Electrophoresis Chambers, Power Supplies, and Accessories Support Center.
Do you have an alternative for the discontinued ZOOM Dual Power Supply?
We offer the PowerEase Touch HV Power Supply as an alternative for the discontinued ZOOM Dual Power Supply.
Find additional tips, troubleshooting help, and resources within our Protein Gel Electrophoresis Chambers, Power Supplies, and Accessories Support Center.
Do I need to use PowerEase Touch HV Power Supply Adapters with the PowerEase Touch HV Power Supply even if I have Invitrogen gel tanks?
The PowerEase Touch HV Power Supply Adapters are designed to facilitate the connection of some electrophoresis tanks to the PowerEase Touch HV Power Supply. The adapters are designed to remain on the electrophoresis tank electrical leads when the lid is removed from the power supply. The supported electrophoresis tanks include:
- ZOOM IPGRunner Mini-cell
- XCell SureLock Mini-Cell
- Mini Gel Tank
- XCell4 SureLock Midi-Cell
Note: The PowerEase Touch HV Power Supply Adapters are not needed for use of the SureLock Tandem Midi Gel Tank with the PowerEase Touch HV Power Supply.
Find additional tips, troubleshooting help, and resources within our Protein Gel Electrophoresis Chambers, Power Supplies, and Accessories Support Center.
Why does the PowerEase Touch HV Power Supply come with power supply adapters?
For safety purposes, the output jacks of the PowerEase Touch HV Power Supply have been recessed compared to other PowerEase Touch power supplies. This requires a firm elongation of the leads on the cables of the tank and this is achieved using the power supply adapters that are provided. The adapters are designed to remain on the electrophoresis tank electrical leads when the lid is removed from the power supply.
Find additional tips, troubleshooting help, and resources within our Protein Gel Electrophoresis Chambers, Power Supplies, and Accessories Support Center.
How many gels can I simultaneously transfer using the PowerEase Touch HV Power Supply?
The PowerEase Touch HV Power Supply can be used for simultaneous transfer of 2 mini gels. We do not recommend using it for transfer of midi gels.
Find additional tips, troubleshooting help, and resources within our Protein Gel Electrophoresis Chambers, Power Supplies, and Accessories Support Center.