Orion™ Pro Star MP218 pH/ion, Conductivity and DO Bench Meter
Orion™ Pro Star MP218 pH/ion, Conductivity and DO Bench Meter
Orion™ Pro Star MP218 pH/ion, Conductivity and DO Bench Meter
Thermo Scientific™

Orion™ Pro Star MP218 pH/ion, Conductivity and DO Bench Meter

Designed to provide accurate and reliable pH, ORP, ion, conductivity, TDS, salinity, resistivity, DO, barometric pressure and temperature analysis, the Thermo Scientific™ Orion™ Pro Star MP218 bench meter is well suited for laboratories that desire a modern meter with advance performance.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoTipo de producto
PSTAR2187Medidor con kit de pH/conductividad
PSTAR2180Medidor con soporte
PSTAR2185Medidor con kit de pH/Cond/OD
PSTAR2186Medidor con kit de pH/Cond/OD óptico
Número de catálogo PSTAR2187
Precio (MXN)
-
Tipo de producto:
Medidor con kit de pH/conductividad

Designed to provide accurate and reliable pH, ORP, ion, conductivity, TDS, salinity, resistivity, DO, barometric pressure and temperature analysis, the Orion Pro Star MP218 bench meter is well suited for labs that desire a modern meter with advance performance. Featuring a 7-in., high-resolution color display with responsive capacitive touchscreen, the Orion Pro Star meter focuses on ensuring an easy to use, innovative meter for your testing needs. The Orion Pro Star meter offers onscreen guidance with step-by-step instructions and supports GLP compliance with date and time stamped logs.

Orion Pro Star MP218 pH/ion, conductivity and dissolved oxygen bench meters are designed to provide accurate and reliable testing in laboratories that need a modern, easy-to-operate meter with advanced performance and reliable data reporting. Orion Pro Star bench meter benefits include:

  • Large 7-in., high-resolution color display
  • Responsive touchscreen operable with gloves
  • Angled display to help ensure ergonomic viewing
  • Streamlined, easy to operate user interface
  • Vertical meter orientation helps optimize bench space
  • Supports GLP compliance
  • Stores 20 calibration logs per parameter with date and time
  • Stores 10,000 data log sets with date and time
  • Outputs to a printer, computer, or USB flash drive for long-term data retention
  • Inputs sample ID with optional USB barcode scanner, USB keyboard or USB mouse
  • Stirs samples with meter-controlled and meter-powered stirrer probes

Orion Pro Star MP218 multiparameter bench meters utilize three channels to offer the following measurements:

  • pH, mV, ORP or ion concentration with temperature
  • Conductivity, TDS, salinity or resistivity with temperature
  • Dissolved oxygen as % saturation or concentration in ppm or mg/L with temperature

First time meter startup is made easy with the meter setup wizard, which guides the user through important meter and measurement settings. Utilize onscreen meter tours and guides for calibration, measurements, meter connections, electrode stand assembly and pH electrode usage to better understand meter operation, warnings and required maintenance. Use guided calibrations with step-by-step instructions to perform calibrations correctly and ensure accurate, reliable and reportable measurements.

Easily view measurements on the large 7-in. color graphic touchscreen display. View measurement channels individually or up to three simultaneously to meet changing measurement needs. Select the basic, advanced or custom measure display format to view only the information you prefer in the measure mode. Select the graph view to display individual measurements in a graph with temperature for measurement versus time studies.

While taking measurements, easily enter sample identification information into the meter using an optional meter accessory including USB barcode scanner, USB keyboard or USB mouse. Easily and uniformly stir samples using an optional meter-controlled and meter-powered stirrer probe, which eliminates the need for a magnetic stir plate and bars.

Save, display and export up to 20 calibrations per parameter with date and time stamp. Filter calibration logs by user and export calibration logs by date range or all at once.

Easily save measurements to the data log for long-term data retention. Save, display and export up to 10,000 data log sets with date and time stamp. View individual data log sets with extensive measurement information organized into an easy-to-read onscreen list format. Filter data logs by user, date and time or log number and export data logs by date and time range, log number range or all at once.

Output meter data logs, calibration logs and methods to a printer, computer or USB flash drive. Save data to a computer or USB drive as comma separated value (CSV) files for import into programs like Microsoft Excel or save data to a USB drive as report files for printable records. Customize exported files to include all data, advanced, intermediate or basic data only to obtain only the information needed for your record keeping. Utilize bidirectional communication with remote commands to interface the meter with computer programs including LIMS, HyperTerminal, PuTTY and additional data collection software.

Save measurement settings to standardize your analysis and SOPs using meter methods. Save up to 10 methods per parameter. Import methods to and export methods from other Orion Pro Star series meters to standardize your testing across multiple meters. Also save, import and export incremental techniques for determination of ion concentration to standardize your known addition, known subtraction, analyte addition and analyte subtraction analyses.

Activate the secure mode and assign up to 3 administrators and up to 30 users with individual account logins and password protection. Administrators can select from pre-defined user access levels to meter functions. Control access to date and time settings, instrument settings, data logs, and calibration logs, or customize each user's access level from full access to basic data collection only. Administrators can also export the user list and audit trail to help ensure digital data integrity.

Operate the included electrode stand to facilitate secure, smooth movement of electrodes into and out of solutions. The electrode stand's weighted base provides flexible placement of the stand around the meter. Secure a wide range of electrodes using the various electrode holder options. Neatly organize electrode cables in the built-in side channels of the stand.

Especificaciones
Adaptador de CAUniversal 100-240 V 50/60 Hz power supply with wall plugs for US/Japan, Euro, UK/Singapore, Australia/New Zealand and China outlets
Exactitud (barómetro)±6.0 mmHg; ±0.008 bar
Exactitud (conductividad)±0.5% reading
Exactitud (DO)±2% saturation; ±0.02 mg/L or ppm
Exactitud (ISE)±0.2 mV or ±0.05% of the reading, whichever is greater
Exactitud (ORP)±0.2 mV or ±0.05% of the reading, whichever is greater
Exactitud (OD polarográfica)±2% saturation; ±0.02 mg/L or ppm
Exactitud (RDO)±2% saturation; ±0.02 mg/L or ppm
Exactitud (resistividad)±0.5% reading
Exactitud (salinidad)±0.5% reading
Exactitud (TDS)±0.5% reading
Exactitud (mV)±0.2 mV or ±0.05% of the reading, whichever is greater
Exactitud (pH)±0.002 pH
Funciones avanzadas de ISESingle known addition, single known subtraction, double known addition, double known subtraction, analyte addition and analyte subtraction
Salida de alarmaCalibration reminder alarm, high and low limit alarm per parameter
Opción de retroiluminaciónSí
Corrección de la presión barométricaYes, automatic with built-in barometer or manual
CalibraciónpH, ORP, Ion Concentration, Conductivity, TDS, Salinity, DO, Barometric Pressure, Temperature
Modos de calibraciónWater-saturated air (100% and 0% saturation), air-saturated water (100% and 0% saturation) or custom concentration value (Winkler)
Puntos de calibración1 to 5 (pH), 1 (ORP), 1 to 5 (ion), 1 to 5 (conductivity), 1 to 5 (TDS), 1 (salinity), 1 to 2 (DO), 1 (barometer), 1 to 2 (temperature)
Puntos de calibración (conductividad)1 to 5 points
Puntos de calibración (ISE)Up to 5
Puntos de calibración (pH)1 to 5 points
Constante de celda0.100 cm-1, 0.475 cm-1, 0.550 cm-1, 1.000 cm-1, 10.00 cm-1 or user-defined from 0.001 to 199.9 cm-1
Certificaciones/conformidadCE, TUV 3-1, FCC Clase A
Canales3
Puntos de datos10,000 with Date and Time Stamp
Dimensiones (Largo x Ancho x Alto)218.9 x 153.2 x 91.33 mm; 8.62 x 6.03 x 3.60 in.
Para utilizar con (aplicación)Most common pH samples as well as pH samples containing tris buffers, proteins, sulfides, halides or other components that can interfere with silver-chloride; wide variety of conductivity samples in the 1 μS/cm to 20 mS/cm range
IncluyeMeter with stand (PSTAR2180); ROSS Ultra Triode refillable epoxy-body pH/ATC electrode (8157BNUMD); 4-cell epoxy/graphite-body K=0.475 conductivity/ATC electrode (013005MD); stirrer probe (096019-WA); pH 4, 7, 10 buffers and rinse solution pouches; pH electrode storage solution and cleaning solution; pH electrode storage sleeve and base; 1413 μS/cm conductivity standard pouches
EntradasChannel 1: BNC for pH, ORP or ISE, 8-pin MiniDIN for ATC, pin-tip for reference; Channel 2: 8-pin MiniDIN for conductivity with ATC; Channel 3: 9-pin MiniDIN for dissolved oxygen with ATC
Punto isopotencial7.000 pH; 1.000 ion concentration
TecladoCapacitive touchscreen user interface
IdiomasChinese, English, French, German, Italian, Japanese, Korean, Portuguese, Spanish
Tipo de función de registroAutomatic with Auto-Read and Timed Read Modes, Manual with Continuous Read Mode
MemoriaNon-volatile
Métodos10 per channel (30 total)
ModeloMP218 Bench Meter pH and Conductivity Kit
SalidasUSB flash drive, computer or printer (STARA-106)
Percent Saturation Range (Polarographic DO)De 0,0 al 600,0% de saturación
Percent Saturation Relative Accuracy (RDO)±2% saturation
Percent Saturation Resolution (Polarographic DO)0,1, 1% de saturación
Intervalo de saturación porcentual (RDO)0.0 to 600.0% saturation
Resolución de saturación en porcentaje (RDO)0,1, 1 % de saturación
Percent Saturation Relative Accuracy (Polarographic DO)±2% saturation
Intervalo de presión400.0 to 850.0 mmHg; 0.533 to 1.133 bar
Tipo de sondaPolarographic or optical DO electrode
Tipo de productoMedidor con kit de pH/conductividad
Intervalo (OD)0.0 to 600.0% saturation; 0.01 to 90.0 mg/L or ppm
Intervalo (ISE)0.0001 to 19999
Intervalo (iones)0.0001 to 19999
Intervalo (ORP)-2000 to 2000 mV
Intervalo (OD polarográfica)0.0 to 600.0% saturation; 0.01 to 90.0 mg/L or ppm
Intervalo (RDO)0.0 to 600.0% saturation; 0.01 to 90.0 mg/L or ppm
Intervalo (mV relativos)-2000 to 2000 mV
Resolución (conductividad)0.001 μS/cm minimum, auto-ranging, up to 4 digits
Resolución (OD)0.1, 1% saturation; 0.01, 0.1 mg/L or ppm
Resolución (ISE)0.0001 minimum, 1 to 5 digits
Resolución (iones)0.0001 minimum, 1 to 5 digits
Resolución (ORP)0.1 mV
Resolución (OD polarográfica)0.1, 1% saturation; 0.01, 0.1 mg/L or ppm
Resolución (RDO)0.1, 1% saturation; 0.01, 0.1 mg/L or ppm
Resolución (resistividad)0.1 Ωminimum, auto-ranging
Resolución (salinidad)0.001 ppt minimum; 0.01 PSU minimum; or 0.01% NaCl minimum, auto-ranging
Resolución (TDS)0.001 ppm or mg/L minimum, auto-ranging, up to 4 digits
Resolución (mV)0.1 mV
Resolución (pH)0.1, 0.01 or 0.001 pH
Factor de salinidad0.0 to 50.0 ppt; automatic with conductivity measurement or manual entry
Modos de medición de salinidadNatural Seawater (ppt), Practical Salinity Units (PSU), or % NaCl
Intervalo de factor de TDSLinear (0.01 to 10.00), ISO/EN 27888, 442, NaCl
Exactitud de temperatura±0,3 °C, ±0,5 °F
Compensación de la temperaturaLinear (0.00 to 10.00 %/°C), nLFn (non-linear natural water), nLFu (non-linear ultra-pure water), EP/USP or off
Temperatura de referencia (métrico)5, 10, 15, 20, 25 or 30°C
Calibración de la sonda de temperatura1 or 2 point temperature offset calibration
Intervalo de temperatura (imperial)-22.0°F to +266.0°F
Resolución de temperatura0,1 °C (0,1 °F)
Selección de temperaturaAutomatic with ATC temperature sensor, ambient or manual
Garantía2 year meter warranty
Peso (imperial)2.20 lbs.
Peso (métrico)1.00 kg
Tipo de pantalla7-in. High-resolution Color Graphic LCD
Clasificación IPIP54
Measurement ModespH, ORP, mV, Ion Concentration (ISE), Conductivity, Dissolved Oxygen (mg/L), Temperature, Barometric Pressure
Intervalo (conductividad)0.010 μS/cm to 3000 mS/cm
Intervalo (resistividad)1.0Ω to 100.0 MΩ
Intervalo (salinidad)0.010 to 80.00 ppt; 0.01 to 42.00 PSU; or 0.01 to 400.0% NaCl
Intervalo (TDS)0.010 ppm to 500.0 ppt (TDS factor of 1.00); or 0.010 mg/L to 500.0 g/L (TDS factor of 1.00)
Intervalo (mV)-2000 - 2000mV
Intervalo (pH)-2 to 20
Intervalo de temperatura (métrico)-30.0°C to +130.0°C
Unit SizeEach