LanthaScreen® Tb-DUB Substrate

Catalog number: PV4443

 Related applications: Pharma & Biopharma | Target & Lead Identification & Validation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Deubiquitinating enzymes (DUBs) play housekeeping functions by maintaining pools of active ubiquitin, and regulatory function by rescuing specific target proteins from degradation by the proteasome. The LanthaScreen® Tb-DUB Substrate is a general substrate for assaying DUB activity in a high throughput assay format.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method: Fluorescent
Excitation⁄Emission (nm): 340⁄490, 520
For Use With (Equipment): Microplate Reader
Form: Frozen
Label or Dye: GFP (EmGFP), Tb (Terbium)
Product Line: LanthaScreen®
Product Size: 10 µg
Readout: Kinetic
Substrate: DUB
Substrate Properties: Protein-Based Substrate
Substrate Type: Deconjugating Enzyme Substrate
Technique: TR-FRET
Shipping Condition: Dry Ice

Contents & storage

LanthaScreen&trade Tb-DUB substate in HEPES buffered saline. Store at -80°C.

Documents

Manuals & protocols