You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Home
▸
Shop All Products
▸
Biochemical Reagents
▸
Peptides
▸
FL-SMRT ID1 Peptide, 100μM
FL-SMRT ID1 Peptide, 100μM
FL-SMRT ID1 PEPTIDE, 100UM
Have Questions?
Catalog Number
Quantity
PV4620
100 μL
Catalog number
PV4620
Price (MXN)
-
Quantity:
100 μL
FL-SMRT ID1 PEPTIDE
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Label Type
Classic Dyes
Quantity
100 μL
Shipping Condition
Dry Ice
Unit Size
Each
Contents & Storage
100 μM Fluorescein-SMRT ID1 Peptide in 99.9% DMSO, 0.1% TFA. Store at -20°C and protect from light.