Product Image

GFP-EIF2A

EIF2A has been expressed as a fusion with GFP (Green Fluorescent Protein) to act as a physiological substrate for theRead more
Have Questions?
Catalog NumberQuantity
PV480920 nmol
Catalog number PV4809
Price (TWD)
-
Quantity:
20 nmol
EIF2A has been expressed as a fusion with GFP (Green Fluorescent Protein) to act as a physiological substrate for the EIF2AK family of kinases including PERK (EIF2AK3) and PKR (EIF2AK2).

For use in combination with the LanthaScreen™ Tb-anti-eIF2alpha [pSer52] Antibody (PV4810) to evaluate inhibitors in medium to high throughput screening applications.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Assay EntryBiochemical terbium phospho-specific activity
Gene ID (Entrez)9833
Label or DyeGFP (EmGFP)
Molecular Weight (g/mol)69100 Da
Quantity20 nmol
Shipping ConditionDry Ice
SubstrateEIF2A
Target EntryEIF2AK1, EIF2AK2, EIF2AK3
Validated KinasesEIF2AK2, EIF2AK3
Detection MethodFluorescence
FormLiquid
Substrate PropertiesProtein-Based Substrate
Substrate TypeKinase Substrate
Target EnzymeSerine/Threonine Kinases
Unit SizeEach
Contents & Storage
GFP-EIF2alpha protein substrate in 9 mM HEPES pH 7.5, 123 mM NaCl, 2.43 mM KCl, 10% Glycerol, 2mM DTT . Store at -80 °C.