Fluorescein-ERM (LRRKtide)
Invitrogen™

Fluorescein-ERM (LRRKtide)

Fluoresceína-ERM (LRRKtido)
Have Questions?
Número de catálogoCantidad
PV49011 mg
Número de catálogo PV4901
Precio (CLP)
-
Cantidad:
1 mg

A fluorescein-labeled peptide substrate based upon ezrin/radixin/moesin (ERM) that is phosphorylated by LRRK2 and LRRK2 mutants.

For use in combination with the LanthaScreen™ Tb-anti-ERM (pLRRKtide) Antibody (PV4899) to evalute inhibitors in medium to high throughput screening applications.

Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Especificaciones
Secuencia de aminoácidosFluoresceína-GAGRLGRDKYKTLRQIRQ
Entrada del ensayoActividad bioquímica fosfoespecífica del terbio
ID de gen (Entrez)120892
Etiqueta o tinteFITC (fluoresceína)
Peso molecular2475,8 Da
Cantidad1 mg
Condiciones de envíoHielo seco
SustratoERM (LRRKtido)
Entrada dianaLRRK2 D1994A, LRRK2 R1441C, LRRK2 I2020T, LRRK2 G2019S, LRRK2
Quinasas validadasLRRK2
Método de detecciónFluorescente
Substrate PropertiesSustrato basado en péptidos
Tipo de sustratoSustrato de cinasa
Target EnzymeSerina/treonina cinasa
Unit SizeEach
Contenido y almacenamiento
400 µM de sustrato peptídico Fluoresceína-ERM (LRRKtido) en agua. Almacenar a -20 °C y proteger de la luz.

Citations & References (1)

Citations & References
Abstract
Investigation of leucine-rich repeat kinase 2 : enzymological properties and novel assays.
Authors:Anand VS, Reichling LJ, Lipinski K, Stochaj W, Duan W, Kelleher K, Pungaliya P, Brown EL, Reinhart PH, Somberg R, Hirst WD, Riddle SM, Steven P B,
Journal:FEBS J
PubMed ID:19076219
Mutations in leucine-rich repeat kinase 2 (LRRK2) comprise the leading cause of autosomal dominant Parkinson's disease, with age of onset and symptoms identical to those of idiopathic forms of the disorder. Several of these pathogenic mutations are thought to affect its kinase activity, so understanding the roles of LRRK2, and ... More