CDK7/9tide Substrate
Thermo Scientific™

CDK7/9tide Substrate

A peptide substrate based upon the C-terminal tail of RNA Polymerase II that is a reported substrate for Cyclin Dependent Kinases (CDK7, CDK8 and CDK9).
Have Questions?
Catalog NumberQuantity
PV50901 mL
Catalog number PV5090
Price (USD)
1,244.00
Each
Quantity:
1 mL
Price (USD)
1,244.00
Each

A peptide substrate based upon the C-terminal tail of RNA Polymerase II that is a reported substrate for Cyclin Dependent Kinases (CDK7, CDK8 and CDK9). For use with the Adapta™ Universal Kinase Assay (Cat. No. PV5099).

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Amino Acid SequenceYSPTSPSYSPTSPSYSPTSPSKKKK
Assay EntryBiochemical ADP accumulation
Gene ID (Entrez)1022, 3376
Molecular Weight (g/mol)2689.9 Da
Quantity1 mL
Shipping ConditionDry Ice
Substratepeptide based upon the C-terminal tail of RNA Polymerase II
Target EntryCDK9/Cyclin T1, CDK7
Validated KinasesCDK7, CDK9
Detection MethodFluorescence
Substrate PropertiesPeptide-Based Substrate
Substrate TypeKinase Substrate
Target EnzymeSerine/Threonine Kinases
Unit SizeEach
Contents & Storage
2 mM CDK7/9tide peptide substrate in 50 mM HEPES (pH 7.5), 0.01% Brij-35, 10 mM MgCl2 and 1 mM EGTA.

Store at -20°C.