Product Image

GFP-STAT1

GFP-STAT1
Have Questions?
Catalog NumberQuantity
PV521110 nmol
Catalog number PV5211
Price (MXN)
-
Quantity:
10 nmol
STAT1 expressed as a fusion with GFP (Green Fluorescent Protein) to act as a physiological substrate for the Janus family of tyrosine kinases.

For use in combination with the LanthaScreen™ Tb-anti-STAT1 [pTyr701] Antibody (PV4844) to evaluate inhibitors in medium to high throughput screening applications.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Assay EntryBiochemical terbium phospho-specific activity
Label or DyeGFP (EmGFP)
Molecular Weight (g/mol)116000 Da
Quantity10 nmol
Shipping ConditionDry Ice
SubstrateSTAT1
Target EntryJAK1, JAK2 JH1 JH2, JAK2, TYK2, JAK3, JAK2 JH1 JH2 V617F
Validated KinasesJAK1, JAK2, JAK3, TYK2
Detection MethodFluorescence
Substrate PropertiesProtein-Based Substrate
Substrate TypeKinase Substrate
Target EnzymeTyrosine Kinases
Unit SizeEach
Contents & Storage
GFP-STAT1 protein substrate in 50 mM Tris-HCl pH 7.5, 100 mM NaCl, 10% Glycerol, 1 mM DTT. Store at -80°C.