Thermo Scientific™

LanthaScreen™ Tb-anti-PRAS40 [pSer183] Antibody Kit

Catalog number: PV5580
Thermo Scientific™

LanthaScreen™ Tb-anti-PRAS40 [pSer183] Antibody Kit

Catalog number: PV5580
Catalog Number
PV5580
Unit Size
25 µg
Price (USD)
3,765.00
Your Price:
Availability
***
Quantity
Catalog Number
Unit Size
1 mg
Price (USD)
17,830.00
Your Price:
Availability
***
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
PV558025 µg
3,765.00
Your Price:
***
PV55811 mg
17,830.00
Your Price:
***
Product Overview
Figures
Videos
Documents
FAQ
Citations & References
Additional Information
Recommendations
Recommendations
The LanthaScreen™ Tb-anti-PRAS40[pSer183] antibody has been designed and validated for use in the LanthaScreen™ PRAS40 HEK 293E Cellular Assay, to facilitate in-house assay configuration and screening. These antibodies may be accessed in small scale and screening quantities.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Concentration
0.5mg/mL, 0.5 mg/ml, 0.5 mg⁄ml, 0.5 mg⁄mL
Host Species
Mouse
Quantity
25 µg
Gene ID (Entrez)
84335
Antibody Conjugate
Lanthanide Chelates, Terbium (Tb)
Antibody Fragment
Whole Antibody
Clonality
Monoclonal
For Use With (Equipment)
Microplate Reader
Gene Symbol
AKT1S1
High-throughput Compatibility
High-throughput Compatible
Host abrv
Ms
Label or Dye
Tb (Terbium)
Label Type
Lanthanide Chelates
Marker
0.5 mg⁄ml
Reactivity
Human
Shipping Condition
Dry Ice
Reactivity abrv
Hu
Sub Class Isotype
IgG
Target Molecule
PRAS40 [pSer183]
Technique
TR-FRET
Product Line
LanthaScreen™

Contents & Storage

Terbium labeled anti-PRAS40[pSer183] antibody in HEPES buffered saline (137 mM NaCl, 2.7 mM KCl, 10 mM HEPES pH 7.5). Storage is at -20 °C

Figures

Frequently asked questions (FAQs)

Citations & References