Product Image

Kinase Tracer 199, 25 μM

In the LanthaScreen® Eu Kinase Binding Assay, binding of an Alexa Fluor® conjugate or 'tracer' such as Kinase Tracer 199Read more
Have Questions?
Catalog NumberQuantity
PV583025 μL
Catalog number PV5830
Price (USD)
670.00
Each
Quantity:
25 μL
Price (USD)
670.00
Each
In the LanthaScreen® Eu Kinase Binding Assay, binding of an Alexa Fluor® conjugate or 'tracer' such as Kinase Tracer 199 to a kinase is detected by addition of a Eu-labeled anti-tag antibody. Binding of the tracer & antibody to a kinase results in a high degree of FRET, whereas displacement of the tracer with a kinase inhibitor results in a loss of FRET. Unlike many kinase activity assays, this assay is a simple mix and read assay, with no development steps. Visit www.invitrogen.com/bindingassay to learn more and to see which kinases have been validated using this kinase tracer.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
AssayKinase Binding Assay
Assay EntryBiochemical binding/kinetic
ColorFar-Red
Excitation/Emission340/665
For Use With (Application)TR-FRET
Label or DyeAlexa Fluor™ 647
Quantity25 μL
Shipping ConditionDry Ice
Target EntryMAPK8, EGFR, RIPK2, MAPK9, EGFR T790M L858R, MAPK14, MAPK10, EGFR L861Q, EGFR L858R
Validated KinasesEGFR T790M L858R, EPHB4, KIT V654A, MAPK9 (JNK2), RIPK2
Detection MethodFluorescence
TypeKinase Tracer
Unit SizeEach
Contents & Storage
Store at -20°C.