Kinase Tracer 222, 50 μM
Thermo Scientific™

Kinase Tracer 222, 50 μM

Kinase Tracer 222 is an Alexa Fluor™ 647 dye-labeled, ATP-competitive kinase inhibitor scaffold for use in the LanthaScreen™ Eu Kinase深入閱讀
Have Questions?
產品號碼Quantity
PV612125 μL
產品號碼 PV6121
價格 (TWD)
19,320.00
線上優惠
Ends: 31-Dec-2026
27,600.00
您節省 8,280.00 (30%)
Each
Quantity:
25 μL
價格 (TWD)
19,320.00
線上優惠
Ends: 31-Dec-2026
27,600.00
您節省 8,280.00 (30%)
Each
Kinase Tracer 222 is an Alexa Fluor™ 647 dye-labeled, ATP-competitive kinase inhibitor scaffold for use in the LanthaScreen™ Eu Kinase Binding Assay.

The LanthaScreen™ Eu Kinase Binding Assay
• Is easily optimized
• Measures inhibitor binding
• Performs simple off-rate measurements

Easily Screen for Kinase Binding
The LanthaScreen™ Eu Kinase Binding Assay is a TR-FRET assay that lets you measure inhibitor binding to non-activated kinases, perform simple off-rate measurements, and makes simple the study of type III (allosteric) inhibitors.

The LanthaScreen™ Eu Kinase Binding Assay is easy to optimize and run. The assay uses a simple mix-and-read format with a single one hour incubation and no development steps. Binding of an Alexa Fluor™ conjugate or “tracer” to a tagged kinase is detected by addition of a europium-labeled anti-tag antibody, in this case anti-GST. Simultaneous binding of the tracer and antibody to the kinase results in a high degree of fluorescence resonance energy transfer (TR-FRET), while displacement of the tracer with a kinase inhibitor results in a loss of TR-FRET.

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
規格
Detection MethodFluorescence
Emission665
Excitation340
For Use With (Application)TR-FRET
For Use With (Equipment)Microplate Reader
Format384-well plate
Label or DyeAlexa Fluor™ 647
Product TypeKinase Tracer 222
Quantity25 μL
ReadoutKinetic
Shipping ConditionDry Ice
TargetKinases (Multiple)
ColorRed
Product LineLanthaScreen
Unit SizeEach
內容物與存放
Store at -20°C