Pre-Weigh MYP (Mannitol Egg Yolk Polymixin) Agar
Imagem ilustrativa. O produto pode variar
Thermo Scientific™

Pre-Weigh MYP (Mannitol Egg Yolk Polymixin) Agar

Speed up your workflow when preparing medium for enumeration of Bacillus cereus in food samples with pre-weighed media sachets.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoQuantity
PW0929-1L20 x Sachets for 1 L Medium
PW0929-8L10 x Sachets for 8 L Medium
Número do catálogo PW0929-1L
Preço (BRL)
-
Quantity:
20 x Sachets for 1 L Medium
Speed up your workflow when preparing microbial culture medium for the enumeration of Bacillus cereus in food samples with Thermo Scientific™ Pre-Weigh MYP (Mannitol Egg Yolk Polymixin) Agar. The pre-weighed sachets of Thermo Scientific™ Oxoid™ dehydrated culture media are designed for single use when preparing 1 L or 8 L volumes of your medium. Save valuable time and start testing quickly by removing the requirement to weigh powder—just rip, tip and pour.

Pre-Weigh MYP Agar is equivalent to the medium developed by Mossel et al.1 It is part of a range of our most frequently used and well-characterized dehydrated culture media which is now available in handy pre-weighed foil packages.

Simply tear open the selected media sachet, add to the required volume of water, mix, and autoclave as usual.

  • Efficient: choose from a full range of pre-weighed media formats for reduced hands-on prep time and maximum productivity
  • Convenient: rip and tip sachets enable faster media production and simplified workflows
  • Compliant: proven and validated media formats deliver confidence and reliable performance
  • Productive: reduce routine dehydrated culture media handling and weighing to spend more time on value-added tasks.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

For Laboratory Use.
For Laboratory Use
References 1. Mossel, D.A.A.; Koopman, M.J. and Jongerius, E. (1967) Appl. Microbiol. 15, 650-653.
Especificações
DescriptionPre-weigh MYP (Mannitol Egg Yolk Polymixin) Agar
Quantity20 x Sachets for 1 L Medium
FormPowder
Product TypeAgar
Unit SizeEach