Qubit™ microRNA Assay Kits
Invitrogen™

Qubit™ microRNA Assay Kits

Achieve accurate miRNA quantitation, even in the presence of rRNA, large mRNA, or contaminants, with the Qubit microRNA Assay Kit, which offers 100 and 500 assay formats.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
Q32881500 Reactions
Q32880100 Reactions
Catalog number Q32881
Price (MXN)
-
No. of Reactions:
500 Reactions
Achieve accurate and precise quantification of small RNA (∼20 nucleotides or base pairs) with Qubit miRNA (microRNA) Assay Kits. These miRNA quantification kits enable quick detection of all types of small RNA, including microRNA and siRNA, and both single stranded and double stranded RNA. The assay is highly selective for small RNA over rRNA or large mRNA, and can tolerate contaminants such as salts, solvents, or detergents.
The Qubit miRNA Assay Kit, when used with the Qubit Fluorometer, provides an accurate and selective method for the quantitation of microRNA samples. This assay is highly selective for miRNA over rRNA or large mRNA (>1000 bp). Depending on sample volume, the assay kit is designed to be accurate for initial miRNA sample concentrations of 0.025 to 150 ng/μL, providing a detection range of 0.5−150 ng.

The Qubit microRNA Assay Kit provides concentrated assay reagent, dilution buffer, and pre-diluted miRNA standards. Simply dilute the reagent using the buffer provided, add your sample (any volume between 1 μL and 20 μL is acceptable), and read the concentration using the Qubit Fluorometer.

Notes
• Use with thin-wall, clear, 0.5-mL PCR tubes (Cat. No. Q32856) for the Qubit 4 Fluorometer and 8 x 200-μL tube strips (Cat. No. Q33252) for the Qubit Flex Fluorometer

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
Excitation/Emission498/518
For Use With (Application)microRNA Quantitation
For Use With (Equipment)Qubit Fluorometer
No. of Reactions500 Reactions
Product LineQubit
Product TypemicroRNA Assay Kit
Quantity500 assays
Shipping ConditionRoom Temperature
Unit SizeEach

Frequently asked questions (FAQs)

How does the accuracy and sensitivity of the Qubit quantitation assays using the Qubit fluorometer compare to a microplate reader?

The accuracy and sensitivity of the Qubit quantitation assays are the same as that of a microplate reader. This was a requirement during product development. The detection limits for each Qubit kit can be found on the corresponding product manual, which can be found by searching our website by keyword or catalog number.

Find additional tips, troubleshooting help, and resources within our Nucleic Acid Quantification Support Center.

Can the Qubit kits give an indication of sample quality in nucleic acid samples?

No. The Qubit DNA and RNA kits only quantify the amount of either DNA or RNA in the sample. The Qubit fluorometer cannot take absorbance readings to provide a A260/A280 ratio or detect protein in nucleic acid samples. This can be done with the NanoDrop instrument. If your sample contains protein or other contaminants that can affect the assay, it should be further purified.

If your sample may contain both DNA and RNA, one may use either (or both) the DNA and RNA Qubit kits and compare with samples treated with either RNase or DNase to get an accurate determination of DNA or RNA, respectively.

Can I use the Quant-iT and Qubit Kits with other fluorometers?

All Quant-iT and Qubit kits are compatible with all fluorometers and microplate readers that have the appropriate light sources and filters. You won't have access to the algorithm in the Qubit fluorometer for generating the standard curve provided by the instrument, instead, you must make a few dilutions of the highest standard DNA or RNA (Standard #2) in the Qubit kits to generate a standard curve with multiple data points.

Citations & References (7)

Citations & References
Abstract
Detection of microRNAs in DNA Extractions for Forensic Biological Source Identification.
Authors:Lewis CA, Layne TR, Seashols-Williams SJ
Journal:J Forensic Sci
PubMed ID:31107550
'Molecular-based approaches for biological source identification are of great interest in the forensic community because of a lack of sensitivity and specificity in current methods. MicroRNAs (miRNAs) have been considered due to their robust nature and tissue specificity; however, analysis requires a separate RNA extraction, requiring an additional step in ... More
Defining quantification methods and optimizing protocols for microarray hybridization of circulating microRNAs.
Authors:Garcia-Elias A, Alloza L, Puigdecanet E, Nonell L, Tajes M, Curado J, Enjuanes C, Díaz O, Bruguera J, Martí-Almor J, Comín-Colet J, Benito B
Journal:Sci Rep
PubMed ID:28798363
MicroRNAs (miRNAs) have emerged as promising biomarkers of disease. Their potential use in clinical practice requires standardized protocols with very low miRNA concentrations, particularly in plasma samples. Here we tested the most appropriate method for miRNA quantification and validated the performance of a hybridization platform using lower amounts of starting ... More
Total RNA extraction from tissues for microRNA and target gene expression analysis: not all kits are created equal.
Authors:Brown RAM, Epis MR, Horsham JL, Kabir TD, Richardson KL, Leedman PJ
Journal:BMC Biotechnol
PubMed ID:29548320
microRNAs (miRNAs) are short non-coding RNAs that fine-tune gene expression. The aberrant expression of miRNAs is associated with many diseases and they have both therapeutic and biomarker potential. However, our understanding of their usefulness is dependent on the tools we have to study them. Previous studies have identified the need ... More
Circulating miRNAs, isomiRs and small RNA clusters in human plasma and breast milk.
Authors:Rubio M, Bustamante M, Hernandez-Ferrer C, Fernandez-Orth D, Pantano L, Sarria Y, Piqué-Borras M, Vellve K, Agramunt S, Carreras R, Estivill X, Gonzalez JR, Mayor A
Journal:PLoS One
PubMed ID:29505615
Circulating small RNAs, including miRNAs but also isomiRs and other RNA species, have the potential to be used as non-invasive biomarkers for communicable and non-communicable diseases. This study aims to characterize and compare small RNA profiles in human biofluids. For this purpose, RNA was extracted from plasma and breast milk ... More
Differences in microRNA expression between melanoma and healthy adjacent skin.
Authors:Aksenenko M, Palkina N, Komina A, Tashireva L, Ruksha T
Journal:BMC Dermatol
PubMed ID:30611259
The tumor microenvironment is composed of cancer-associated fibroblasts, tumor-associated macrophages, endothelial cells, immune cells, signaling molecules and extracellular matrix structures, which closelycommunicate with the tumor via multiple mechanisms. MicroRNAs are paracrine regulators that provide a direct interaction between the microenvironment and cancer cells. In the presentstudy, we aimed to identify ... More