Qubit™ Flex Endotoxin Detection Starter Kit with Standard Warranty - FAQs
Qubit™ Flex Endotoxin Detection Starter Kit with Standard Warranty - FAQs
View additional product information for Qubit™ Flex Endotoxin Detection Starter Kit with Warranty - FAQs (Q32894REXPKG, Q32894, Q32894PKG)
10 product FAQs found
Does the Qubit Flex NGS Starter Kit (Cat. No. Q45893) include the Qubit Flex System Verification Assay Kit (Cat. No. Q33254)?
No. The Qubit Flex NGS Starter Kit (Cat. No. Q45893) does not include the Qubit Flex System Verification Assay Kit (Cat. No. Q33254). It must be purchased separately.
The description of the Qubit Flex Starter Packages (Cat. Nos. Q32894, Q45893, Q45894) can be found on the Qubit Flex Starter Packages product page.
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Quantification Support Center.
Does the Qubit Flex Quantitation Starter Kit (Cat. No. Q45894) include a Qubit dsDNA Broad Range Assay Kit (Cat. No. Q33265, Q33266)?
No. The Qubit Flex Quantitation Starter Kit (Cat. No. Q45894) does not include a Qubit dsDNA Broad Range Assay Kit (Cat. No. Q33265, Q33266). It must be purchased separately.
The description of the Qubit Flex Starter Packages (Cat. Nos. Q32894, Q45893, Q45894) can be found on the Qubit Flex Starter Packages product page.
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Quantification Support Center.
Can I use the Qubit Endotoxin Detection Assay Kit on the Qubit 4 Fluorometer?
No, the Qubit Endotoxin Detection Assay Kit is designed for use with the Qubit Flex Fluorometer and is not currently for use on the Qubit 4 Fluorometer.
To use the Qubit Endotoxin Detection Assay Kit on the Qubit Flex Fluorometer, do I need to perform a software update ?
Yes, the endotoxin detection assay program will be available on Qubit Flex Software version 1.8.0 or later. The latest software is available under the Resources section of www.thermofisher.com/qubit
How do I store the Qubit Endotoxin Detection Assay Kit?
The entire kit can be stored at 2-8 degrees C for 6 months. Reconstituted Endotoxin standard solutions are stable for 4 weeks at 2-8 degrees C and reconstituted dye can be stored for 4 weeks at 2-8 degrees C. Reconstituted amebocyte lysate solution is stable for 1 week at -20 degrees C or colder if frozen immediately after reconstitution.
Is the Qubit Flex Fluorometer (Cat. No. Q33327) compatible with custom assays?
No, the Qubit Flex Fluorometer does not have custom assay capability.
The settings options on my Qubit instrument are greyed out and I am being prompted to sign in. How can I resolve this?
Our Qubit 4 and Qubit Flex instruments will now require users to sign in to access certain menu options (i.e., instrument settings, software update, and system verification). This is necessary to adhere to cybersecurity legal standards that were put in place in 2020. Only when signed in, are all functions of the device are accessible.
Instructions for login:
- Turn on the device
- From the home screen, touch the person icon on the top left corner
- Touch the "Get Started" button
- Touch the "Create Profile" button
- Enter information for username and pin
- Log in by touching the person icon and entering the user info in the sign in fields
Which is the best DNA or RNA quantification instrument?
All of the DNA instruments offered by Thermo Fisher Scientific are of excellent quality. To find the right instrument to quantify DNA or RNA in your lab, explore the detailed comparison of UV-Vis spectrophotometers and fluorometers. Things like sensitivity, throughput and budget may be initial considerations when selecting a DNA quantification instrument. You may also consider whether you need target specificity, sample purity information, RNA quality information, or broad dynamic range.
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Quantification Support Center.
What is the difference between UV and fluorescence DNA quantification? Which DNA quantification technology should I choose?
UV and fluorescence technologies work differently to quantify DNA. UV quantification relies on the intrinsic absorptivity of DNA and RNA molecules, while fluorescence quantification uses dyes that specifically bind to your molecule of choice. With UV technology, quantification isn't as sensitive, but it has a broader dynamic range and also gives data about sample purity‒plus it is faster because there is no reagent prep. With fluorescence technology you get higher sensitivity and molecule-specific data, but it has lower dynamic range and reagent prep is required. The technology you need will depend on what features are important for your lab and experimentation.
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Quantification Support Center.
When using the Qubit Flex Fluorometer, do I have to use all 8 samples/wells?
No, the Qubit Flex Fluorometer allows you to select which wells will be used for the experiment. The standards and sample will only be read in the selected wells. If the option to select wells is not available on your Qubit Flex, please update to the current firmware version (download firmware from Technical Resources for Qubit Fluorometers). This feature was introduced in version 1.5.0.