New—The Qubit™ 4 Fluorometer can now measure RNA quality in addition to providing accurate quantitation of dsDNA, ssDNA, RNA & protein.
Qubit™ Protein and Protein Broad Range (BR) Assay Kits
Invitrogen™

Qubit™ Protein and Protein Broad Range (BR) Assay Kits

Achieve sensitive detection of protein, and low protein-to-protein variation, with the Qubit Protein and Protein BR Assay Kits for fluorescence-based protein quantitation assays.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberAssayQuantitation RangeSufficient For
Q33212Qubit Protein Assay12.5 μg/mL to 5 mg/mL500 Reactions
Q33211Qubit Protein Assay12.5 μg/mL to 5 mg/mL100 Reactions
A50668Qubit Protein Broad Range Assay0.1 to 20 mg/mL100 Reactions
A50669Qubit Protein Broad Range Assay0.1 to 20 mg/mL500 Reactions
Catalog number Q33212
Price (USD)
471.65
Online exclusive
502.00
Save 30.35
Each
Assay:
Qubit Protein Assay
Quantitation Range:
12.5 μg/mL to 5 mg/mL
Sufficient For:
500 Reactions
Price (USD)
471.65
Online exclusive
502.00
Save 30.35
Each
Quantify protein quickly and accurately with two unique Qubit protein quantification kits: the Qubit Protein Assay, which quantifies 12.5 μg/mL to 5 mg/mL of protein, and the Qubit Protein BR Assay Kit, which quantifies 0.1 to 20 mg/mL of protein. Both assays are compatible with many common contaminants, reducing reagents, salts, and nucleic acids. The Protein BR Assay Kit is also tolerant of detergents.

Qubit Protein Assay Kit
The Qubit Protein Assay Kit is designed specifically for use with any Qubit fluorometer. While using between 1 and 20 μL of sample, this assay can quantify samples ranging from 12.5 μg/mL to 5 mg/mL with low protein-to-protein variation.

The Qubit Protein Assay Kit is highly selective for proteins and is also highly accurate in the presence of reducing reagents, but not in the presence of large amounts of detergent. Common contaminants, such as reducing reagents (e.g., DTT, β-mercaptoethanol), salts, free nucleotides, amino acids, solvents, and DNA (but not detergents), are well tolerated in the assay. Slight protocol modifications are required for other contaminants.

The kit provides concentrated assay reagent, dilution buffer, and pre-diluted BSA standards. Simply dilute the reagent using the buffer provided, add your sample (any volume between 1 μL and 20 μL is acceptable), and read the concentration.

Which product should you choose for fluorometric protein quantitation?
• 1–20 samples: use this Qubit Protein Assay Kit with the Qubit Fluorometer
• 20–2000 samples: use the Quant-iT Protein Assay Kit with a microplate reader

Qubit Protein BR Assay Kit
The Qubit Protein BR Assay Kit provides a quick, easy, and reliable method for determining the protein concentration of purified proteins, lysates, serum, plasma, or complex protein mixtures using a Qubit 4 Fluorometer. This protein kit can deliver accurate protein quantitation across a broad range (0.1–20 mg/mL), enabling most samples to be used neat (undiluted) and eliminating the guess work and dilution steps that accompany traditional protein quantitation methods. The assay is compatible with many common contaminants, such as detergents, reducing reagents (e.g., DTT, β-mercaptoethanol), salts, and nucleic acids.

Key features of the Qubit Protein BR Assay Kit include:
• Broad dynamic range—0.1 to 20 mg/mL, measure samples without dilution
• Two-point standard curve—eliminate time-consuming standard preparation
• Automatic concentration determination—receive protein concentration instantaneously; no need for offline calculations
• Compatible—accurate in the presence of reducing reagents, detergents, and other common buffer components

The Qubit Protein BR Assay Kit comes with two calibration standards, Protein BR Assay Buffer, and Protein BR Assay Reagent. Simply add 150–160 μL of Protein BR Assay Buffer to 10 or 20 μL of your sample, followed by 30 μL of Protein BR Assay Reagent. Incubate for 10 minutes at room temperature. Then insert the sample into a Qubit 4 instrument and read the concentration.

Notes
• The Qubit Protein Assay Kit can be used with the Qubit 1.0, Qubit 2.0, Qubit 3, Qubit 4, and Qubit Flex fluorometers.
• The Qubit Protein BR Assay Kit is a fluorescence-based assay intended for use only with the Qubit 4 Fluorometer
• 500 μL thin-walled clear PCR tubes (Cat. No. Q32856) are required but not included in the Protein Assay and BR Assay Kits

For Research Use Only. Not for use in diagnostic procedures.
Specifications
AssayQubit Protein Assay
For Use With (Equipment)Qubit Fluorometer, Microplate Reader
Product LineQubit
Product TypeProtein Quantification Assay
Quantitation Range12.5 μg/mL to 5 mg/mL
Quantity500 Assays
Shipping ConditionRoom Temperature
Sufficient For500 Reactions
Detection MethodFluorescence
Unit SizeEach

Frequently asked questions (FAQs)

How does the accuracy and sensitivity of the Qubit quantitation assays using the Qubit fluorometer compare to a microplate reader?

The accuracy and sensitivity of the Qubit quantitation assays are the same as that of a microplate reader. This was a requirement during product development. The detection limits for each Qubit kit can be found on the corresponding product manual, which can be found by searching our website by keyword or catalog number.

Find additional tips, troubleshooting help, and resources within our Nucleic Acid Quantification Support Center.

Can the Qubit kits give an indication of sample quality in nucleic acid samples?

No. The Qubit DNA and RNA kits only quantify the amount of either DNA or RNA in the sample. The Qubit fluorometer cannot take absorbance readings to provide a A260/A280 ratio or detect protein in nucleic acid samples. This can be done with the NanoDrop instrument. If your sample contains protein or other contaminants that can affect the assay, it should be further purified.

If your sample may contain both DNA and RNA, one may use either (or both) the DNA and RNA Qubit kits and compare with samples treated with either RNase or DNase to get an accurate determination of DNA or RNA, respectively.

Can I use the Quant-iT and Qubit Kits with other fluorometers?

All Quant-iT and Qubit kits are compatible with all fluorometers and microplate readers that have the appropriate light sources and filters. You won't have access to the algorithm in the Qubit fluorometer for generating the standard curve provided by the instrument, instead, you must make a few dilutions of the highest standard DNA or RNA (Standard #2) in the Qubit kits to generate a standard curve with multiple data points.

Can I use the original Quant-iT Kits with the Qubit Fluorometer?

No, we do not recommend this. Some of the dyes in the original Quant-iT kits (those NOT listed as “for use with the Qubit fluorometer”) are not compatible with the Qubit Fluorometer. In addition, the new Quant-iT kits (labeled as “for use with the Qubit Fluorometer”) have standards formulated to be compatible with the Qubit fluorometer internal algorithms for the respective assays. The Qubit Fluorometer-compatible kits are also less expensive per assay if you are processing fewer than 20 samples at a time.

Is 1X RIPA buffer (50 mM Tris-HCl, 150 mM NaCl, 0.25% deoxycholic acid, 1% NP-40, 1mM EDTA, pH 7.4) compatible with the Qubit Protein Assay (Cat. No. Q33211/Q33212)?

The concentration of detergents in 1x RIPA buffer is quite high, therefore, we do not recommend that you use this kit with samples in RIPA. The Qubit Protein Assay has a tolerance for detergents present only in very low amounts. Please see the user guide for the tolerance of this assay for detergents here. The Qubit Protein Broad Range (BR) Assay Kit (A50668) has a much higher tolerance for detergents, e.g. RIPA buffer can be used undiluted. Please see this link.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Quantitation Support Center.

Citations & References (23)

Citations & References
Abstract
Post-training disruption of Arc protein expression in the anterior cingulate cortex impairs long-term memory for inhibitory avoidance training.
Authors:Holloway CM, McIntyre CK,
Journal:Neurobiol Learn Mem
PubMed ID:21315825
'The activity-regulated-cytoskeletal-associated protein (Arc) has a well established role in memory consolidation and synaptic plasticity in the hippocampus and amygdala. However the role of Arc within the anterior cingulate cortex (ACC), an area of the brain involved in processing memory for pain, has yet to be examined. Here we sought ... More
Investigation of the involvement of P-glycoprotein and multidrug resistance-associated protein 2 in the efflux of ximelagatran and its metabolites by using short hairpin RNA knockdown in Caco-2 cells.
Authors:Darnell M, Karlsson JE, Owen A, Hidalgo IJ, Li J, Zhang W, Andersson TB,
Journal:Drug Metab Dispos
PubMed ID:20023051
'Liver and bile secretion can be an important first-pass and clearance route for drug compounds and also the site of several drug-drug interactions. In the clinical program for ximelagatran development, an unexpected effect of erythromycin on the pharmacokinetics of the direct thrombin inhibitor ximelagatran and its metabolites was detected. This ... More
Synthesis and optimization of lectin functionalized nanoprobes for the selective recovery of glycoproteins from human body fluids.
Authors:Ferreira JA, Daniel-da-Silva AL, Alves RM, Duarte D, Vieira I, Santos LL, Vitorino R, Amado F,
Journal:Anal Chem
PubMed ID:21809823
'Biomedical sciences, and in particular biomarker research, demand efficient glycoprotein enrichment platforms. Herein magnetic nanoprobes (MNP), after being coated with three broad-spectrum lectins-concanavalin A (ConA), wheat germ agglutinin (WGA), and Maackia amurensis lectin (MA)-were utilized to selectively capture glycoproteins from human body fluids. Additionally, a new methodology, based on protection ... More
Active and total transforming growth factor-ß1 are differentially regulated by dopamine and estradiol in the pituitary.
Authors:Recouvreux MV, Guida MC, Rifkin DB, Becu-Villalobos D, Díaz-Torga G,
Journal:Endocrinology
PubMed ID:21521749
'Dopamine, acting through the dopamine type 2 receptor (Drd2), is the main inhibitor of pituitary prolactin (PRL) secretion and lactotroph proliferation. TGF-ß1 is involved, at least in part, in mediating these actions. It was described that TGF-ß1 synthesis in rat pituitary lactotrophs is up-regulated by dopamine and down-regulated by estradiol. ... More
Silencing of NHE-1 blunts the slow force response to myocardial stretch.
Authors:Pérez NG, Nolly MB, Roldan MC, Villa-Abrille MC, Cingolani E, Portiansky EL, Alvarez BV, Ennis IL, Cingolani HE,
Journal:J Appl Physiol (1985)
PubMed ID:21659487
'Myocardial stretch induces a biphasic force response: a first abrupt increase followed by a slow force response (SFR), believed to be the in vitro manifestation of the Anrep effect. The SFR is due to an increase in Ca²? transient of unclear mechanism. We proposed that Na?/H? exchanger (NHE-1) activation is ... More