Use 96- and 384-well Microplates for Fluorescence-based Assays with Quant-iT assays for optimal results
Quant-iT™ RNA Assay Kits and Quant-iT RNA HS Reagent
Invitrogen™

Quant-iT™ RNA Assay Kits and Quant-iT RNA HS Reagent

Achieve highly selective and accurate quantification of high-abundance RNA samples with the fluorescent Quant-iT RNA Reagent and Assay Kits. These kits are highly selective for RNA over double-stranded DNA, and provide sufficient material for at least 2000 assays in a 96-well microplate format.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityAssayQuantitation Range
Q332251000 assaysRNA Quantitation, ext range200 to 10,000 ng
Q331401 kitRNA Quantitation5 to 100 ng
Q102131,000 assaysRNA Quantitation, BR20 to 1000 ng
Q328841 mLRNA Quantitation, HS5 to 100 ng
Catalog number Q33225
Price (USD)
600.65
Online exclusive
646.00
Save 45.35
Each
Quantity:
1000 assays
Assay:
RNA Quantitation, ext range
Quantitation Range:
200 to 10,000 ng
Price (USD)
600.65
Online exclusive
646.00
Save 45.35
Each
Enable easy and accurate RNA quantitation with the Quant-iT RNA Assay Kit, which is also available in broad range (BR) and extended range (XR) configurations, as well as a stand-alone high sensitivity (HS) reagent. These Quant-iT RNA quantification kits are highly selective for RNA over dsDNA and can be used to generate fluorescence-based linear detection ranges of RNA.
Quant-iT RNA Reagent and Assay Kit
The Quant-iT RNA Reagent and Assay Kit makes RNA quantification easy and accurate. The kit provides concentrated assay reagent, dilution buffer, and pre-diluted RNA standards. The assay is highly selective for RNA over double-stranded DNA, and provides a dynamic range of 5-100 ng of RNA.

Quant-iT RNA BR (Broad Range) Assay Kit
The Quant-iT RNA BR (Broad Range) Assay Kit provides an accurate and selective method for quantification of high-abundance RNA samples. While the original Quant-iT RNA kit has great sensitivity, the Quant-iT RNA Broad Range kit is ideal for microarray samples where the concentration of RNA is higher. The assay is highly selective for RNA over double-stranded DNA, and provides a dynamic range of 20-1000 ng of RNA.

Quant-iT RNA XR (Extended Range) Assay Kit
The Quant-iT RNA XR (Extended Range) Assay Kit provides an accurate and selective method for the quantification of high-abundance RNA samples. The assay is highly selective for RNA and does not quantitate DNA, protein, or free nucleotides. The assay kit is designed to be accurate for initial RNA sample concentrations of 10 ng/μL to 10,000 ng/μL depending on sample volume, providing a linear detection range of 200–10,000 ng.

For each assay, simply dilute the reagent using the buffer provided, add the sample (any volume between 1 μL and 20 μL is acceptable), and read the concentration using a fluorescence-based microplate reader. Common contaminants, such as salts, free nucleotides, solvents, detergents, and protein, are well tolerated.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
AssayRNA Quantitation, ext range
For Use With (Equipment)Microplate Reader
Product LineQuant-iT
Quantitation Range200 to 10,000 ng
Quantity1000 assays
Shipping ConditionDry Ice
Detection MethodFluorescence
Unit SizeEach

Frequently asked questions (FAQs)

Why am I getting negative fluorescence values with my Qubit Assays?

Negative fluorescence is a physical impossibility. It is an artifact from software autocorrecting for background signal. This means your reader is picking up and subtracting out background light at the cost of your data. Make sure to do a buffer-only control and assess the type of signal. You may need to switch to a different plate.

Citations & References (2)

Citations & References
Abstract
The tumor inflammation signature (TIS) is associated with anti-PD-1 treatment benefit in the CERTIM pan-cancer cohort.
Authors:Damotte D, Warren S, Arrondeau J, Boudou-Rouquette P, Mansuet-Lupo A, Biton J, Ouakrim H, Alifano M, Gervais C, Bellesoeur A, Kramkimel N, Tlemsani C, Burroni B, Duche A, Letourneur F, Si H, Halpin R, Creasy T, Herbst R, Ren X, Morel P, Cesano A, Goldwasser F, Leroy K
Journal:J Transl Med
PubMed ID:31684954
The 18-gene tumor inflammation signature (TIS) is a clinical research assay that enriches for clinical benefit to immune checkpoint blockade. We evaluated its ability to predict clinical benefit of immunotherapy in cancer patients treated with PD-1 checkpoint inhibitors in routine clinical care. ... More
Detection and expression analysis of
Authors:Arredondo A, Àlvarez G, Nart J, Mor C, Blanc V, León R
Journal:J Oral Microbiol
PubMed ID:31448060
Tetracycline resistance can be achieved through