Qubit™ 1X dsDNA High Sensitivity (HS) and Broad Range (BR) Assay Kits
Invitrogen™

Qubit™ 1X dsDNA High Sensitivity (HS) and Broad Range (BR) Assay Kits

Highly selective for dsDNA over RNA, the Qubit 1X dsDNA HS and BR Assay Kits offer easy-to-use formulations for the accurate quantitation of dsDNA in 100 and 500 assay formats. Lambda Standards provide sufficient material for up to 500 samples.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityQuantitation Range
Q33231500 assays0.1 to 120 ng
Q33230100 assays0.1 to 120 ng
Q33266500 assays4 to 4000 ng
Q33265100 assays4 to 4000 ng
Catalog number Q33231
Price (USD)
462.65
Online exclusive
487.00
Save 24.35
Each
Quantity:
500 assays
Quantitation Range:
0.1 to 120 ng
Price (USD)
462.65
Online exclusive
487.00
Save 24.35
Each
Experience easy, accurate, and precise quantification of dsDNA with Qubit 1X dsDNA HS (High Sensitivity) and Qubit 1X dsDNA BR (Broad Range) Assay Kits. The 1X formulation provides a ready-to-use working solution that enables simple dsDNA sample quantification on Qubit Fluorometers. These assays are highly selective for dsDNA over ssDNA, RNA, protein, and free nucleotides. Contaminants, such as salts, solvents, or detergents are well-tolerated.
Qubit 1X dsDNA HS and BR Assay Kits, designed for use with Qubit Fluorometers, are highly selective for double-stranded DNA (dsDNA) over single-stranded DNA (ssDNA), RNA, protein, and free nucleotides. All kits provide a ready-to-use working solution and DNA standards. To perform the assay, simply dilute your sample (any volume from 1–20 μL is acceptable) into the 1X working solution provided, then read the concentration using a Qubit Fluorometer.

Qubit 1X dsDNA HS Assay Kit
The Qubit dsDNA HS (High Sensitivity) Assay Kit, when used with the Qubit Fluorometer, provides an accurate and selective method for the quantitation of sensitive DNA samples. Depending on sample volume, the assay kit is designed to be accurate for initial DNA sample concentrations of 5 pg/μL to 120 ng/μL, providing a detection range of 0.1−120 ng.

Qubit 1X dsDNA BR Assay Kit
The Qubit dsDNA BR (Broad-Range) Assay Kit, designed for use with Qubit 4 or Qubit Flex Fluorometers, provides an accurate and selective method for the quantitation of DNA samples. Depending on sample volume, the assay kit is designed to be accurate for initial DNA sample concentrations of 0.2 to 4,000 ng/μL, providing a detection range of 4-4,000 ng.

Notes
• Qubit 1X dsDNA HS can be used with Qubit 2, Qubit 3, Qubit 4, and Qubit Flex Fluorometers
• Qubit 1X dsDNA BR are designed for use with Qubit 4 and Qubit Flex Fluorometers
• Use thin-wall, clear, 0.5-mL PCR tubes (Cat. No. Q32856) for the Qubit 4 Fluorometer and 8 x 200 μL tube strips (Cat. No. Q33252) for the Qubit Flex Fluorometer

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Assay1X dsDNA, high sensitivity
For Use With (Equipment)Qubit 2.0, 3, 4, and Flex Fluorometers
No. of Reactions500 Reactions
Product LineQubit
Quantitation Range0.1 to 120 ng
Quantity500 assays
Shipping ConditionRoom Temperature
Unit SizeEach

Frequently asked questions (FAQs)

What is the expiration date for Qubit 1X dsDNA High Sensitivity (HS) and Broad Range (BR) Assay Kits (Cat. Nos. Cat. Q33230, Q33231, Q33265, Q33266)?

The Qubit 1X dsDNA High Sensitivity (HS) and Broad Range (BR) Assay Kits (Cat. Nos. Cat. Q33230, Q33231, Q33265, Q33266) are stable for 6 months from the date of receipt when stored as directed.

For more information on product storage, please refer to the user guides:

Qubit 1X dsDNA HS Assay Kit and Qubit 1X dsDNA BR Assay Kit

Find additional tips, troubleshooting help, and resources within our Nucleic Acid Quantification Support Center.

What is the difference between The Qubit 1X dsDNA High Sensitivity (HS) and Broad Range (BR) Assay Kits (Cat. Nos. Q33230, Q33231, Q33265, Q33366) and Qubit dsDNA Quantification Assay Kits (Cat. Nos. Q32850, Q32851, Q32853, Q32854)?

The Qubit 1X dsDNA High Sensitivity (HS) and Broad Range (BR) Assay Kits (Cat. Nos. Q33230, Q33231, Q33265, Q33366) are newer and have a simplified workflow compared to the original Qubit dsDNA Quantification Assay Kits (Cat. Nos. Q32850, Q32851, Q32853, Q32854). The original Qubit kits contain separate dyes and buffer components that must be mixed together to make a working solution, in which the dye degrades after a few hours. With the newer Qubit 1X dsDNA Assay kits, the dye is premixed with a buffer that keeps the dye stable long-term and can be added directly to DNA samples.

You can find more information about the Qubit 1X dsDNA assay, by clicking on the Technical Note provided below:

Qubit 1X dsDNA assays: simplified workflow and improved performance

Find additional tips, troubleshooting help, and resources within our Nucleic Acid Quantification Support Center.

How should I store my Qubit 1X dsDNA High Sensitivity (HS) and Broad Range (BR) Assay Kits (Cat. No. Q33230, Q33231, Q33265, Q33266)?

The Qubit 1X dsDNA High Sensitivity (HS) (Cat. Nos. Q33230, Q33231) and Broad Range (BR) Assay Kits (Cat. Nos. Q33265, Q33266) have different storage conditions.

For Qubit 1X dsDNA HS Assay Kits (Cat. Nos. Q33230, Q33231), store all components at 2 - 8 degrees C.

For Qubit 1X dsDNA BR Assay Kits (Cat. Nos. Q33265, Q33266), store the working solution at 18 - 28 degrees C. Store the standards at 2 - 8 degrees C.

The assay kits are stable for at least 6 months from the date of receipt.

For more information, please refer to these user guides: Qubit 1X dsDNA HS Assay Kit and Qubit 1X dsDNA BR Assay Kit

Find additional tips, troubleshooting help, and resources within our Nucleic Acid Quantification Support Center.

Citations & References (7)

Citations & References
Abstract
Plasmodium-specific atypical memory B cells are short-lived activated B cells.
Authors:Pérez-Mazliah D, Gardner PJ, Schweighoffer E, McLaughlin S, Hosking C, Tumwine I, Davis RS, Potocnik AJ, Tybulewicz VL, Langhorne J,
Journal:Elife
PubMed ID:30387712
'A subset of atypical memory B cells accumulates in malaria and several infections, autoimmune disorders and aging in both humans and mice. It has been suggested these cells are exhausted long-lived memory B cells, and their accumulation may contribute to poor acquisition of long-lasting immunity to certain chronic infections, such ... More
Co-digestion of organic and mineral wastes for enhanced biogas production: Reactor performance and evolution of microbial community and function.
Authors:Shamurad B, Gray N, Petropoulos E, Tabraiz S, Acharya K, Quintela-Baluja M, Sallis P
Journal:Waste Manag
PubMed ID:31109531
'Mineral wastes (MWs) from municipal solid waste incineration plants and construction demolition sites are rich in minerals, heavy metals and have acid neutralising capacity. This renders such MWs a promising source of bulk and trace elements to enhance and stabilize biogas production in anaerobic processes. However, finding a MW with ... More
Unravelling the Role of Trophoblastic-Derived Extracellular Vesicles in Regulatory T Cell Differentiation.
Authors:Kovács ÁF, Fekete N, Turiák L, Ács A, Kohidai L, Buzás EI, Pállinger É
Journal:Int J Mol Sci
PubMed ID:31337116
'Regulatory T cells (T'
Genomic Epidemiology of SARS-CoV-2 in Guangdong Province, China.
Authors:Lu J, du Plessis L, Liu Z, Hill V, Kang M, Lin H, Sun J, François S, Kraemer MUG, Faria NR, McCrone JT, Peng J, Xiong Q, Yuan R, Zeng L, Zhou P, Liang C, Yi L, Liu J, Xiao J, Hu J, Liu T, Ma W, Li W, Su J, Zheng H, Peng B, Fang S, Su W, Li K, Sun R, Bai R, Tang X, Liang M, Quick J, Song T, Rambaut A, Loman N, Raghwani J, Pybus OG, Ke C
Journal:Cell
PubMed ID:32359424
'Coronavirus disease 2019 (COVID-19) is caused by SARS-CoV-2 infection and was first reported in central China in December 2019. Extensive molecular surveillance in Guangdong, China''s most populous province, during early 2020 resulted in 1,388 reported RNA-positive cases from 1.6 million tests. In order to understand the molecular epidemiology and genetic ... More
High-throughput isolation of giant viruses using high-content screening.
Authors:Francis R, Ominami Y, Bou Khalil JY, La Scola B,
Journal:Commun Biol
PubMed ID:31240254
The race to discover and isolate giant viruses began 15 years ago. Metagenomics is counterbalancing coculture, with the detection of giant virus genomes becoming faster as sequencing technologies develop. Since the discovery of giant viruses, many efforts have been made to improve methods for coculturing amebas and giant viruses, which ... More