Qubit NGS Library Quantification Assay Kit
Qubit NGS Library Quantification Assay Kit
Qubit NGS Library Quantification Assay Kit
Qubit NGS Library Quantification Assay Kit
Invitrogen™

Qubit NGS Library Quantification Assay Kit

The Invitrogen™ Qubit™ NGS Library Quantification Assay is an easy-to-use fluorescence-based assay for precise molar quantification of sequencing-ready Illumina next-generation sequencing (NGS) libraries using the Qubit™ Flex Fluorometer or compatible fluorescence microplate readers.
Have Questions?
Catalog NumberQuantity
Q342501 Ea.
Catalog number Q34250
Price (USD)
290.00
Each
Quantity:
1 Ea.
Price (USD)
290.00
Each

The Invitrogen™ Qubit™ NGS Library Quantification Assay is an easy-to-use fluorescence-based assay for precise molar quantification of sequencing-ready Illumina next-generation sequencing (NGS) libraries using the Qubit™ Flex Fluorometer or compatible fluorescence microplate readers. The assay selectively quantifies P5/P7-adapted double-stranded DNA libraries prior to sequencing, helping ensure optimal cluster density and successful sequencing runs.

Accurate determination of library concentration is a critical step in NGS workflows, as the amount of DNA loaded onto a flow cell directly impacts cluster density, data quality, and sequencing efficiency. The Qubit NGS Library Quantification Assay provides a dilution-free, time-saving alternative to qPCR-based library quantification methods while delivering comparable precision.

Key features and benefits:

  • Selective molar quantification—measures only dual-adapted, sequencing-ready libraries for improved accuracy
  • Dilution-free workflow—eliminates error-prone serial dilution steps required for qPCR
  • Broad dynamic range—quantifies libraries from 0.8–46 nM without additional dilution
  • Robust performance—accurately quantifies target libraries even in the presence of contaminating nucleic acids
  • Flexible detection—compatible with the Qubit Flex Fluorometer and fluorescence microplate readers
  • Accessible and cost-effective—no need for dedicated qPCR instrumentation

The streamlined workflow reduces hands-on time and minimizes potential user error, enabling efficient library quantification for both genomic core facilities and individual laboratories. By delivering precise quantification of sequencing-ready libraries, the Qubit NGS Library Quantification Assay helps ensure optimal cluster density on flow cells, improving sequencing outcomes.

Notes

  • Designed for use with the Qubit Flex Fluorometer (Cat. No. Q33327) and fluorescence-based microplate readers. The assay is not currently available for use on Qubit 4 Fluorometers.
  • Optimized for use with Qubit Flex Pyrogen Free Assay Tube Strips (Cat. No. Q32893).
  • Requires the use of the Qubit Flex System Verification Assay Kit (Cat. No. Q33254) prior to the first assay run.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
Emission515 nm
Excitation490 nm
No. of Reactions96 Reactions
Product TypeFluorescence Assay
Quantity1 Ea.
FormatKit
Isolation TechnologyMagnetic Bead
Sequencing TypeNext Generation Sequencing
Unit SizeEach