QuantiGene™ Plex Mouse Neuroscience Panel, 15-plex
Invitrogen™

QuantiGene™ Plex Mouse Neuroscience Panel, 15-plex

The QuantiGene Plex Mouse Neuroscience Panel 15-plex, enables measurement of expression of relevant mouse genes associated with neuroscience and brain development through the analysis of 15 RNA targets and housekeeping genes in a single well using Luminex xMAP technology.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
QGP-315-MSNEUROSCI10 Plates
QGP-115-MSNEUROSCI1 Plate
QGP-215-MSNEUROSCI3 Plates
Catalog number QGP-315-MSNEUROSCI
Price (USD)
23,040.00
Each
Quantity:
10 Plates
Price (USD)
23,040.00
Each

The QuantiGene Plex Mouse Neuroscience Panel, 15-plex, enables measurement of expression of relevant genes in mice associated with neuroscience and brain development through the analysis of 15 RNA targets and housekeeping genes in a single well using Luminex xMAP technology in combination with branched DNA signal amplification. This panel contains key genes that are involved in dynamic processes regulating brain development and neurobiology.

Targets: Ahsg, Apoe, Bdnf, Cfh, Cntf, Fgf21, Gdnf, Gfap, Ncam1, Ngf, Nrgn, Uchl1, Gapdh, Hprt, Ppib

QuantiGene Plex panels are extensively tested for target combinability, interference, and cross-reactivity to provide outstanding levels of validation and precision.

This panel is used in combination with the QuantiGene Plex Assay Kit, which contains all buffers, plates, and detection reagents necessary to perform the assay in 96-well plates.

Features of QuantiGene Plex panels include:

  • True multiplexing—measure multiple genes of interest and housekeeping genes in the same well with no cross-reactivity
  • Accommodation of difficult sample types—works with degraded and cross-linked RNA in FFPE tissues and blood
  • Standardized platform—uses 96-plate format compatible with Luminex 200, FLEXMAP 3D, and INTELLIFLEX systems (384-well format available on demand)
  • Simple workflow—employs an ELISA-like workflow with direct hybridization of transcripts to beads and transcript labeling (branched DNA signal amplification and PE readout)
  • Customize with validated targets—if alternative targets are needed in this panel, select from our large inventory of over 22,000 genes to create custom research panels
  • Rapid turnaround for fast delivery

Panel and signal amplification technology

QuantiGene Plex assays are hybridization-based assays for the direct quantitation of RNA directly from cell lysates, tissue homogenates, or purified RNA, and they enable the measurement of multiple targets in the same well via multiplexing. QuantiGene Plex panels contain two components: a target-specific probe set and magnetic capture beads. The target-specific probes are used to capture the RNA of interest, and branched DNA technology then amplifies the fluorescent signals, which are read with a Luminex xMAP instrument. The intensity of the fluorescent signal is directly proportional to the amount of target mRNA present in the sample. This allows for (relative) quantification of gene expression and, with this specific panel, enables human immune profiling. Detailed and step-by-step data analysis is facilitated by the QuantiGene Plex data analysis app.

Applications

QuantiGene Plex assay research applications include compound screening of drug candidates, biomarker verification, siRNA knockdown efficiency determination, prospective and/or retrospective analysis of clinical samples, microarray validation, predictive toxicology research, and translocation/fusion gene detection. In general, any multiplex gene expression analysis can be done with QuantiGene Plex assays, independent of species or sample type.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)QuantiGene Gene Expression Assay
For Use With (Equipment)Luminex Instrument
Product LineQuantiGene™ Plex
Sample TypePurified RNA, cell/blood/FFPE lysates, tissue homogenates
Shipping ConditionWet Ice
Detection MethodFluorescence
Format96-well
Product TypeMultiplex Panel
Quantity10 Plates
Research AreaNeurobiology
SpeciesMouse
Unit SizeEach
Contents & Storage
Target-specific Probe Set and magnetic Capture Beads (premixed and ready to use)
Store Probe Set at -20°C and Capture Beads at 4°C (do not freeze!)