QuantiGene™ Plex Mouse Immune Response Panel, 80-plex
Invitrogen™

QuantiGene™ Plex Mouse Immune Response Panel, 80-plex

The QuantiGene Plex Mouse Immune Response Panel, 80-plex, enables the study of immune function in mice by analyzing 80 RNA targets in a single well using Luminex xMAP technology in combination with branched DNA signal amplification.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수량
QGP-380-2006410 Plates
QGP-180-200641 Plate
QGP-280-200643 Plates
카탈로그 번호 QGP-380-20064
제품 가격(KRW)
110,174,000
Each
수량:
10 Plates
제품 가격(KRW)
110,174,000
Each

The QuantiGene Plex Mouse Immune Response Panel, 80-plex enables the study of immune function in mice by analyzing 80 RNA targets in a single well using Luminex xMAP technology in combination with branched DNA signal amplification. Analyze 80 cytokine, chemokine, and growth factor targets simultaneously for efficient immune response profiling, biomarker discovery, and validation.

QuantiGene Plex panels are extensively tested for target combinability, interference, and cross-reactivity to provide the highest level of validation and precision. This kit is not compatible with the MAGPIX instrument.

This panel is used in combination with the QuantiGene Plex Assay Kit (Cat. No. QP1013), which contains all buffers, plates, and detection reagents necessary to perform the assay in 96-well plates.

Features of the QuantiGene Plex Mouse Immune Response Panel, 80-plex include:

  • True multiplexing—measure 80 genes of interest and housekeeping genes in the same well with no cross-reactivity
  • Accommodation of difficult sample types—works with degraded and cross-linked RNA in FFPE tissues and blood
  • Standardized platform—uses 96-well plate format compatible with Luminex 200, FLEXMAP 3D, and INTELLIFLEX systems (384-well format available on demand)
  • Simple workflow—employs an ELISA-like workflow with direct hybridization of transcripts to beads and transcript labeling (branched DNA signal amplification and PE readout)
  • Customize with validated targets—if alternative targets are needed in this panel, select from our large inventory of over 22,000 genes to create custom research panels
  • Rapid turnaround for this catalog panel for fast delivery

Targets (80-plex): Il2ra, Il2, Il6, Ifng, Tnf, Il5, Il1a, Csf2, Il4, Il10, Il18, Vegfa, Il17a, Il1b, Il12a, Ccl2, Ccl7, Ccl11, Ccl5, Cxcl11, Il6ra, Ccl3, Ccl4, Il13, Il23a, Cxcr3, Il22, Il15, Il27, Il33, Ifna1, Il28a, Il31, Cxcl1, Cxcl2, Csf3, Il3, Lep, Tnfsf11, Csf1, Lif, Il9, Btc, Cxcl5, Il25, Il1rl1, Il19, Cd27, Kdr, Ccl19, Cxcl16, Il16, Ccl22, Ccl12, Gzmb, Ccl27a, Ccl24, Ccl17, Ccl25, Cxcl13, Il7r, Il7, Tslp, Tnfsf13b, Il12b, Il21, Gzma, Cd274, Ctla4, Cxcl9, Havcr2, Lag3, Ifna2, Ifnb1, Hgf, Tnfrsf12a, Ppib, Hprt, Gapdh, Gusb

Panel and signal amplification technology

QuantiGene Plex assays are hybridization-based assays for the direct quantitation of RNA from cell lysates, tissue homogenates, or purified RNA. They enable the measurement of multiple targets in the same well via multiplexing.

QuantiGene Plex panels contain two components: a target-specific probe set and magnetic capture beads. The target-specific probes are used to capture the RNA of interest, and branched DNA technology then amplifies the fluorescent signals, which are read with a Luminex xMAP instrument. The fluorescent signal is directly proportional to the amount of target mRNA present in the sample. This allows for (relative) quantification of gene expression and with this specific panel enables human immune profiling. Detailed and step-by-step data analysis is facilitated by the QuantiGene Plex data analysis app.

Applications

QuantiGene Plex assay research applications include compound screening of drug candidates, biomarker verification, siRNA knockdown efficiency, prospective and/or retrospective analysis of clinical samples, microarray validation, predictive toxicology, and detection of translocations and fusion genes. In general, any multiplex gene expression analysis can be done with QuantiGene Plex assays, independent of species or sample type.

For Research Use Only. Not for use in diagnostic procedures.
사양
용도(애플리케이션)QuantiGene Gene Expression Assay
용도(장비)Luminex Instrument
제품라인QuantiGene™ Plex
샘플 종류Purified RNA, cell/blood/FFPE lysates, tissue homogenates
배송 조건Wet Ice
검출 방법Fluorescence
형식96-well
제품 유형Multiplex Panel
수량10 Plates
Research AreaImmunology
Mouse
Unit SizeEach
구성 및 보관

Target-specific probe set and magnetic capture beads (premixed and ready to use).

Store probe set and capture beads at 4°C.

Do not freeze the capture beads, as they can be damaged if frozen.