Invitrogen™

QuantiGene™ Lysis Mixture

Catalog number: QP0523
Invitrogen™

QuantiGene™ Lysis Mixture

Catalog number: QP0523
Catalog Number
QP0523
Unit Size
36 mL
Price (USD)
Price: 177.00
Your price:
Availability
-
Quantity
Catalog Number
Unit Size
12 mL
Price (USD)
Price: 74.50
Your price:
Availability
-
Quantity
Catalog Number
Unit Size
120 mL
Price (USD)
Price: 442.00
Your price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
QP052336 mL
Price: 177.00
Your price:
-
QP052212 mL
Price: 74.50
Your price:
-
QP0524120 mL
Price: 442.00
Your price:
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

QuantiGene Lysis Mixture is used to lyse cells from cell culture or whole blood, in order to release target RNA to be quantified with QuantiGene assays. QuantiGene Lysis Mixture is one of two components in the QuantiGene Sample Processing kits for cells and whole blood, and is intended to supplement these kits when additional volume is required.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Application)
Multi-Gene Expression
Product Line
QuantiGene™
Product Type
Cell Lysis Buffer
Quantity
36 mL
Shipping Condition
Room Temperature

Contents & Storage

Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text