Invitrogen™

QuantiGene™ Lysis Mixture

Catalog number: QP0524
Invitrogen™

QuantiGene™ Lysis Mixture

Catalog number: QP0524
Catalog Number
QP0524
Unit Size
120 mL
Price (USD)
375.00
Your Price:
Availability
***
Quantity
Catalog Number
Unit Size
12 mL
Price (USD)
62.75
Your Price:
Availability
***
Quantity
Catalog Number
Unit Size
36 mL
Price (USD)
150.00
Your Price:
Availability
***
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
QP0524120 mL
375.00
Your Price:
***
QP052212 mL
62.75
Your Price:
***
QP052336 mL
150.00
Your Price:
***
Product Overview
Figures
Videos
Documents
FAQ
Citations & References
Additional Information
Recommendations
Recommendations
QuantiGene Lysis Mixture is used to lyse cells from cell culture or whole blood, in order to release target RNA to be quantified with QuantiGene assays. QuantiGene Lysis Mixture is one of two components in the QuantiGene Sample Processing kits for cells and whole blood, and is intended to supplement these kits when additional volume is required.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Application)
Multi-Gene Expression
Product Line
QuantiGene™
Product Type
Cell Lysis Buffer
Shipping Condition
Blue Ice

Contents & Storage

Store at room temperature.

Figures

Frequently asked questions (FAQs)

Citations & References