QuantiGene™ Plex Magnetic Separation Plates
Invitrogen™
QuantiGene™ Plex Magnetic Separation Plates
QuantiGene Magnetic Separation Plates are 96-well flat-bottom plates used for the branched DNA and SAPE incubations on the second day of the QuantiGene Plex Assay.
Have Questions?
Catalog NumberQuantity
QP054010 Plates
Catalog number QP0540
Price (MXN)
-
Quantity:
10 Plates
QuantiGene Magnetic Separation Plates are 96-well flat-bottom plates used for the branched DNA and SAPE incubations on the second day of the QuantiGene Plex Assay. The Magnetic Separation Plates are compatible with the Hand-Held Magnetic Plate Washer (Cat. No. EPX-55555-000) and QuantiGene Plex Plate Seals (Cat. No. QG0524). These plates are one component of several included in the QuantiGene Plex Assay kit and are offered here as a supplement to the assay kit when more plates are needed.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeMagnetic Separation Plate
Quantity10 Plates
Shipping ConditionRoom Temperature
Array8 x 12
BarcodeNo Barcode
No. of Wells96
Product LineQuantiGene Plex
Well ShapeFlat Bottom (F-well)
Unit SizeEach