Invitrogen™
Handheld Magnetic Plate Washer
Catalog number:  QP0702
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
The Handheld Magnetic Plate Washer is used with QuantiGene Plex assays or ProcartaPlex assays for all magnetic bead washing steps. The washer enables users to remove the liquid contents of a 96-well plate while retaining the beads.
How it works
The Handheld Magnetic Plate Washer contains 96 magnets that pull and hold magnetic beads on the bottom of each well, allowing users to simply flick the wash buffer into a sink and blot the remainder on a paper towel.
What you get
The magnetic frame is white polycarbonate with a corrosion resistant plate seal below, both set on a polypropylene base. The clip system provided is capable of holding a variety of microplates. The ring on the base plate facilitates gripping for all sizes of hands.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product Size: 1 washer
Shipping Condition: Room Temperature
Documents
Manuals & protocols