QuantiGene™ Plex Wash Buffer Components
Invitrogen™

QuantiGene™ Plex Wash Buffer Components

QuantiGene Plex Wash Buffer Components are aqueous buffered solutions that are mixed with nuclease-free water to make QuantiGene Plex wash buffer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
QP150310 Plates
QP15011 Plate
QP15023 Plates
Catalog number QP1503
Price (EUR)
323,00
Each
Quantity:
10 Plates
Price (EUR)
323,00
Each
QuantiGene Plex Wash Buffer Components are aqueous buffered solutions that are mixed with nuclease-free water to make QuantiGene Plex wash buffer. The wash buffer components may be purchased in 1, 3, or 10-plate quantities to supplement QuantiGene Plex Assay kits when additional wash buffer volume is required for automation.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)QuantiGene Plex Assay
Key FunctionsMulti-Gene Expression
Product LineQuantiGene Plex
Shipping ConditionRoom Temperature
Format96-well
Product TypeWash Buffer
Quantity10 Plates
Unit SizeEach
Contents & Storage
• 8 mL Wash Buffer Component 1
• 130 mL Wash Buffer Component 2

Store at room temperature.